Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582735_m1
          See other WFS1 GE Assays ›
          Gene Symbol:
          Wfs1
          Gene Name:
          Wolfram syndrome 1 (wolframin)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 78640707 - 78665224 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582735_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wfs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031823.1 NP_114011.1 1 - 2 254 95
          GenBank mRNA AF136378.1 - 1 - 2 254 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000787.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wfs1

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          renal water homeostasis
          visual perception
          sensory perception of sound
          ER-associated ubiquitin-dependent protein catabolic process
          endoplasmic reticulum unfolded protein response
          pancreas development
          positive regulation of protein ubiquitination
          endoplasmic reticulum calcium ion homeostasis
          response to endoplasmic reticulum stress
          olfactory behavior
          glucose homeostasis
          negative regulation of programmed cell death
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of neuron apoptotic process
          positive regulation of proteolysis
          positive regulation of growth
          protein stabilization
          neurological system process
          positive regulation of protein metabolic process
          positive regulation of calcium ion transport
          calcium ion homeostasis
          polyubiquitinated misfolded protein transport
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          negative regulation of type B pancreatic cell apoptotic process
          transporter activity
          calmodulin binding
          ubiquitin protein ligase binding
          activating transcription factor binding
          calcium-dependent protein binding
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P440558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171266 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139433 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P440558 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P440558 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P440558 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P440558 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700636987 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102831 - -
          Agilent Rat cDNA Microarray (G4105A) 700636987 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773292 - -
          Affymetrix Rat Gene 1.0 ST Array 10773293 - -
          Affymetrix Rat Gene 1.0 ST Array 10773294 - -
          Affymetrix Rat Gene 1.0 ST Array 10773296 - -
          Affymetrix Rat Gene 1.0 ST Array 10773291 - -
          Affymetrix Rat Gene 1.0 ST Array 10773297 - -
          Affymetrix Rat Gene 1.0 ST Array 10773295 - -
          Affymetrix Rat Gene 1.1 ST Array 10773290 - -
          Rat Genome 230 2.0 Array 1368839_at - -
          Rat Genome 230 2.0 Array 1368839_PM_at - -
          Rat Genome 230 2.0 Array 1387356_PM_at - -
          Rat Genome 230 2.0 Array 1387356_at - -
          Affymetrix HT Rat Focus Array 1368839_PM_at - -
          Affymetrix HT Rat Focus Array 1387356_at - -
          Affymetrix HT Rat Focus Array 1387356_PM_at - -
          Affymetrix HT Rat Focus Array 1368839_at - -
          Affymetrix Rat Clariom S Assay PSR1400005382.rn.1 TC1400000787.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400005385.rn.1 TC1400000787.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400005383.rn.1 TC1400000787.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400005384.rn.1 TC1400000787.rn.1 RTA TC
          Rat Expression Set 230 1368839_PM_at - -
          Rat Expression Set 230 1368839_at - -
          Rat Expression Set 230 1387356_at - -
          Rat Expression Set 230 1387356_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773290 - -
          Rat Genome U34 Set rc_AI172032_at - -
          Rat Genome U34 Set rc_AI230698_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21494523 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358741 - -
          RatRef-12 Expression BeadChip ILMN_54749 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline