Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582903_m1
          See other RTN4 GE Assays ›
          Gene Symbol:
          Rtn4
          Gene Name:
          reticulon 4
          Gene Aliases:
          NI-250, Nogo, Nogo-A, Vp20, r, rat N, rat NogoA
          Chromosome Location:
          Chr.14: 114123517 - 114174459 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582903_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rtn4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031831.1 NP_114019.1 5 - 6 3527 90
          XM_006251608.1 XP_006251670.1 3 - 4 1304 90
          XM_006251609.2 XP_006251671.1 3 - 4 1014 90
          XM_008770468.1 XP_008768690.1 4 - 5 5114 90
          GenBank mRNA AF051335.1 - 3 - 4 439 90
          AF132045.1 - 4 - 5 1623 90
          AF132046.1 - 3 - 4 1103 90
          AJ242961.1 - 5 - 6 3527 90
          AJ242962.1 - 3 - 4 1054 90
          AJ242963.1 - 3 - 4 500 90
          AY164740.1 - 3 - 4 1054 90
          AY164741.1 - 4 - 5 1623 90
          BC070879.1 - 3 - 4 1239 90
          BC097936.1 - 3 - 4 1059 90
          BC161841.1 - 3 - 4 1060 90
          FQ213608.1 - 1 - 2 99 90
          FQ228713.1 - 2 - 3 237 90
          FQ235341.1 - 2 - 3 200 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001194.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rtn4

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          nervous system development
          axonal fasciculation
          aging
          negative regulation of neuron projection development
          response to activity
          olfactory nerve development
          cerebral cortex radial glia guided migration
          negative regulation of cell growth
          regulation of cell migration
          negative regulation of axon extension
          positive regulation of dopamine secretion
          negative regulation of neuron differentiation
          positive regulation of glial cell differentiation
          oligodendrocyte differentiation
          negative regulation of axonogenesis
          regulation of sensory perception of pain
          regulation of nervous system development
          cardiac epithelial to mesenchymal transition
          negative regulation of neuron projection regeneration
          endoplasmic reticulum tubular network organization
          regulation of branching morphogenesis of a nerve
          protein binding
          protein complex binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P132516 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P477322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101531 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59242 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20702 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143497 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132516 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P477322 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132516 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P477322 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P132516 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P477322 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P477322 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700539965 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92470 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69964 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14627 - -
          Agilent Rat cDNA Microarray (G4105A) 700539965 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10774767 - -
          Affymetrix Rat Gene 1.0 ST Array 10774774 - -
          Affymetrix Rat Gene 1.0 ST Array 10774771 - -
          Affymetrix Rat Gene 1.0 ST Array 10774781 - -
          Affymetrix Rat Gene 1.0 ST Array 10774772 - -
          Affymetrix Rat Gene 1.0 ST Array 10774780 - -
          Affymetrix Rat Gene 1.0 ST Array 10774778 - -
          Affymetrix Rat Gene 1.0 ST Array 10774776 - -
          Affymetrix Rat Gene 1.0 ST Array 10774768 - -
          Affymetrix Rat Gene 1.0 ST Array 10774777 - -
          Affymetrix Rat Gene 1.0 ST Array 10774779 - -
          Affymetrix Rat Gene 1.0 ST Array 10774769 - -
          Affymetrix Rat Gene 1.0 ST Array 10774775 - -
          Affymetrix Rat Gene 1.1 ST Array 10774766 - -
          Rat Genome 230 2.0 Array 1368888_PM_a_at - -
          Rat Genome 230 2.0 Array 1368888_a_at - -
          Rat Genome 230 2.0 Array 1388027_a_at - -
          Rat Genome 230 2.0 Array 1388027_PM_a_at - -
          Affymetrix HT Rat Focus Array 1388027_PM_a_at - -
          Affymetrix HT Rat Focus Array 1388027_a_at - -
          Affymetrix HT Rat Focus Array 1368888_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368888_a_at - -
          Affymetrix Rat Clariom S Assay PSR1400008749.rn.1 TC1400001194.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400008753.rn.1 TC1400001194.rn.1 RTA TC
          Rat Expression Set 230 1368888_a_at - -
          Rat Expression Set 230 1388027_a_at - -
          Rat Expression Set 230 1388027_PM_a_at - -
          Rat Expression Set 230 1368888_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10774766 - -
          Rat Genome U34 Set rc_AI105064_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21986321 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356186 - -
          RatRef-12 Expression BeadChip ILMN_64945 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline