Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582990_m1
          See other NDRG4 GE Assays ›
          Gene Symbol:
          Ndrg4
          Gene Name:
          NDRG family member 4
          Gene Aliases:
          Bdm1, Ndr4, smap8
          Chromosome Location:
          Chr.19: 9766439 - 9802387 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582990_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndrg4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271091.1 NP_001258020.1 7 - 8 637 62
          NM_001271092.1 NP_001258021.1 7 - 8 637 62
          NM_001271093.1 NP_001258022.1 7 - 8 584 62
          NM_001271094.1 NP_001258023.1 7 - 8 584 62
          NM_001271095.1 NP_001258024.1 5 - 6 502 62
          NM_031967.2 NP_114173.2 5 - 6 502 62
          XM_006255123.2 XP_006255185.1 7 - 8 579 62
          GenBank mRNA AF045564.1 - 5 - 6 434 62
          AY217029.1 - 5 - 6 473 62
          AY217030.1 - 7 - 8 584 62
          AY217031.1 - 7 - 8 584 62
          AY217032.1 - 7 - 8 632 62
          AY217033.1 - 7 - 8 632 62
          AY255791.1 - 5 - 6 473 62
          CB585874.1 - 1 - 2 59 62
          FM054107.1 - 5 - 6 455 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000970.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndrg4

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          brain development
          visual learning
          negative regulation of platelet-derived growth factor receptor signaling pathway
          positive regulation of neuron projection development
          negative regulation of smooth muscle cell migration
          vesicle docking
          negative regulation of smooth muscle cell proliferation
          cell migration involved in heart development
          positive regulation of ERK1 and ERK2 cascade
          regulation of endocytic recycling
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P034654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222578 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46438 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130449 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P034654 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P034654 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P034654 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12703 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65694 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38485 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48088 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22654 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10809117 - -
          Affymetrix Rat Gene 1.0 ST Array 10809120 - -
          Affymetrix Rat Gene 1.0 ST Array 10809111 - -
          Affymetrix Rat Gene 1.0 ST Array 10809103 - -
          Affymetrix Rat Gene 1.0 ST Array 10809114 - -
          Affymetrix Rat Gene 1.0 ST Array 10809118 - -
          Affymetrix Rat Gene 1.0 ST Array 10809105 - -
          Affymetrix Rat Gene 1.0 ST Array 10809119 - -
          Affymetrix Rat Gene 1.0 ST Array 10809113 - -
          Affymetrix Rat Gene 1.0 ST Array 10809107 - -
          Affymetrix Rat Gene 1.0 ST Array 10809109 - -
          Affymetrix Rat Gene 1.0 ST Array 10809106 - -
          Affymetrix Rat Gene 1.0 ST Array 10809101 - -
          Affymetrix Rat Gene 1.0 ST Array 10809115 - -
          Affymetrix Rat Gene 1.0 ST Array 10809104 - -
          Affymetrix Rat Gene 1.0 ST Array 10809108 - -
          Affymetrix Rat Gene 1.0 ST Array 10809110 - -
          Affymetrix Rat Gene 1.0 ST Array 10809102 - -
          Affymetrix Rat Gene 1.1 ST Array 10809100 - -
          Rat Genome 230 2.0 Array 1370229_PM_at - -
          Rat Genome 230 2.0 Array 1370229_at - -
          Affymetrix HT Rat Focus Array 1370229_at - -
          Affymetrix HT Rat Focus Array 1370229_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900007588.rn.1 TC1900000970.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007589.rn.1 TC1900000970.rn.1 RTA TC
          Rat Expression Set 230 1370229_PM_at - -
          Rat Expression Set 230 1370229_at - -
          Affymetrix Rat Gene 1.0 ST Array 10809100 - -
          Rat Genome U34 Set AF045564_at - -
          Rat Genome U34 Set AF045564_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21979282 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365550 - -
          RatRef-12 Expression BeadChip ILMN_67987 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline