Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583276_m1
          See other DLL1 GE Assays ›
          Gene Symbol:
          Dll1
          Gene Name:
          delta-like 1 (Drosophila)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 57318621 - 57326732 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583276_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dll1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032063.2 NP_114452.1 4 - 5 955 84
          GenBank mRNA U78889.2 - 4 - 5 955 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004572.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dll1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate determination
          somitogenesis
          somite specification
          heart looping
          Notch signaling pathway
          cell-cell signaling
          determination of left/right symmetry
          compartment pattern specification
          nervous system development
          auditory receptor cell fate commitment
          negative regulation of interleukin-10 production
          negative regulation of cell differentiation
          negative regulation of auditory receptor cell differentiation
          negative regulation of myeloid cell differentiation
          negative regulation of neuron differentiation
          positive regulation of Notch signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          inner ear development
          regulation of neurogenesis
          left/right axis specification
          proximal tubule development
          loop of Henle development
          neuronal stem cell population maintenance
          Notch binding
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P130025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659866 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122835 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130025 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130025 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P130025 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P423921 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89459 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718284 - -
          Affymetrix Rat Gene 1.0 ST Array 10718287 - -
          Affymetrix Rat Gene 1.0 ST Array 10718290 - -
          Affymetrix Rat Gene 1.0 ST Array 10718293 - -
          Affymetrix Rat Gene 1.0 ST Array 10718288 - -
          Affymetrix Rat Gene 1.0 ST Array 10718292 - -
          Affymetrix Rat Gene 1.0 ST Array 10718294 - -
          Affymetrix Rat Gene 1.0 ST Array 10718291 - -
          Affymetrix Rat Gene 1.0 ST Array 10718286 - -
          Affymetrix Rat Gene 1.0 ST Array 10718289 - -
          Affymetrix Rat Gene 1.0 ST Array 10718295 - -
          Affymetrix Rat Gene 1.0 ST Array 10718285 - -
          Affymetrix Rat Gene 1.1 ST Array 10718283 - -
          Rat Genome 230 2.0 Array 1368938_at - -
          Rat Genome 230 2.0 Array 1368938_PM_at - -
          Affymetrix HT Rat Focus Array 1368938_at - -
          Affymetrix HT Rat Focus Array 1368938_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100036203.rn.1 TC0100004572.rn.1 RTA TC
          Rat Expression Set 230 1368938_at - -
          Rat Expression Set 230 1368938_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10718283 - -
          Rat Genome U34 Set rc_AI136950_s_at - -
          Rat Genome U34 Set U78889_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21037454 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363310 - -
          RatRef-12 Expression BeadChip ILMN_49221 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline