Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583283_m1
          See other SLC1A6 GE Assays ›
          Gene Symbol:
          Slc1a6
          Gene Name:
          solute carrier family 1 (high affinity aspartate/glutamate transporter), member 6
          Gene Aliases:
          EAAT4
          Chromosome Location:
          Chr.7: 13730101 - 13758274 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583283_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc1a6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032065.1 NP_114454.1 2 - 3 347 68
          XM_008765182.1 XP_008763404.1 2 - 3 521 68
          GenBank mRNA AY286330.1 - 2 - 3 341 68
          BC098912.1 - 2 - 3 371 7315936 68
          U89608.1 - 2 - 3 347 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002223.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc1a6

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          L-glutamate transport
          regulation of membrane potential
          L-glutamate transmembrane transport
          anion transmembrane transport
          L-glutamate transmembrane transporter activity
          anion transmembrane transporter activity
          sodium:dicarboxylate symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122742 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P523524 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P523524 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P523524 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P523524 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88036 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69248 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900881 - -
          Affymetrix Rat Gene 1.0 ST Array 10900883 - -
          Affymetrix Rat Gene 1.0 ST Array 10900884 - -
          Affymetrix Rat Gene 1.0 ST Array 10900877 - -
          Affymetrix Rat Gene 1.0 ST Array 10900876 - -
          Affymetrix Rat Gene 1.0 ST Array 10900875 - -
          Affymetrix Rat Gene 1.0 ST Array 10900874 - -
          Affymetrix Rat Gene 1.0 ST Array 10900880 - -
          Affymetrix Rat Gene 1.0 ST Array 10900882 - -
          Affymetrix Rat Gene 1.0 ST Array 10900878 - -
          Affymetrix Rat Gene 1.0 ST Array 10900885 - -
          Affymetrix Rat Gene 1.0 ST Array 10900879 - -
          Affymetrix Rat Gene 1.1 ST Array 10900873 - -
          Rat Genome 230 2.0 Array 1387161_at - -
          Rat Genome 230 2.0 Array 1387161_PM_at - -
          Affymetrix HT Rat Focus Array 1387161_PM_at - -
          Affymetrix HT Rat Focus Array 1387161_at - -
          Affymetrix Rat Clariom S Assay PSR0700017780.rn.1 TC0700002223.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700017787.rn.1 TC0700002223.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700017786.rn.1 TC0700002223.rn.1 RTA TC
          Rat Expression Set 230 1387161_PM_at - -
          Rat Expression Set 230 1387161_at - -
          Affymetrix Rat Gene 1.0 ST Array 10900873 - -
          Rat Genome U34 Set U89608_at - -
          Rat Genome U34 Set U80915_s_at - -
          Rat Genome U34 Set U80915_s_at - -
          Rat Genome U34 Set U89608_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20988804 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351486 - -
          RatRef-12 Expression BeadChip ILMN_69392 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline