Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583376_m1
          See other KCNQ1 GE Assays ›
          Gene Symbol:
          Kcnq1
          Gene Name:
          potassium channel, voltage-gated KQT-like subfamily Q, member 1
          Gene Aliases:
          Kvlqt1
          Chromosome Location:
          Chr.1: 216293087 - 216630339 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583376_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnq1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032073.1 NP_114462.1 11 - 12 1607 85
          GenBank mRNA AJ133685.1 - 11 - 12 1607 85
          AY043394.1 - 10 - 11 1347 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002934.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnq1

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of gene expression by genetic imprinting
          potassium ion transport
          male gonad development
          positive regulation of heart rate
          gene silencing
          regulation of ion transmembrane transport
          cellular response to drug
          regulation of membrane potential
          negative regulation of insulin secretion
          cardiac muscle contraction
          regulation of membrane repolarization
          regulation of ventricular cardiac muscle cell membrane repolarization
          regulation of atrial cardiac muscle cell membrane repolarization
          positive regulation of cardiac muscle contraction
          positive regulation of gastric acid secretion
          cellular response to cAMP
          potassium ion export
          potassium ion transmembrane transport
          response to anesthetic
          cardiovascular system development
          ventricular cardiac muscle cell action potential
          membrane repolarization during action potential
          membrane repolarization during cardiac muscle cell action potential
          atrial cardiac muscle cell action potential
          regulation of heart rate by cardiac conduction
          potassium ion export across plasma membrane
          positive regulation of potassium ion transmembrane transport
          voltage-gated potassium channel activity
          delayed rectifier potassium channel activity
          potassium channel activity
          protein binding
          calmodulin binding
          protein phosphatase 1 binding
          outward rectifier potassium channel activity
          protein kinase A catalytic subunit binding
          protein kinase A regulatory subunit binding
          protein homodimerization activity
          ion channel binding
          voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization
          voltage-gated potassium channel activity involved in atrial cardiac muscle cell action potential repolarization
          scaffold protein binding
          voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P787920 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P094313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43840 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112502 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149182 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104082 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100787 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73742 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138134 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34455 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131366 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59181 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20361 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2904 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094313 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P787920 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P787920 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094313 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P094313 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P787920 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P212703 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23290 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13414 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25101 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69978 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22292 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53065 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64358 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65662 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71551 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48949 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57778 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91650 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10712539 - -
          Affymetrix Rat Gene 1.0 ST Array 10712532 - -
          Affymetrix Rat Gene 1.0 ST Array 10712535 - -
          Affymetrix Rat Gene 1.0 ST Array 10712542 - -
          Affymetrix Rat Gene 1.0 ST Array 10712541 - -
          Affymetrix Rat Gene 1.0 ST Array 10712544 - -
          Affymetrix Rat Gene 1.0 ST Array 10712543 - -
          Affymetrix Rat Gene 1.0 ST Array 10712547 - -
          Affymetrix Rat Gene 1.0 ST Array 10712534 - -
          Affymetrix Rat Gene 1.0 ST Array 10712531 - -
          Affymetrix Rat Gene 1.0 ST Array 10712545 - -
          Affymetrix Rat Gene 1.0 ST Array 10712537 - -
          Affymetrix Rat Gene 1.0 ST Array 10712546 - -
          Affymetrix Rat Gene 1.0 ST Array 10712536 - -
          Affymetrix Rat Gene 1.0 ST Array 10712533 - -
          Affymetrix Rat Gene 1.0 ST Array 10712540 - -
          Affymetrix Rat Gene 1.0 ST Array 10712538 - -
          Affymetrix Rat Gene 1.1 ST Array 10712530 - -
          Rat Genome 230 2.0 Array 1368371_at - -
          Rat Genome 230 2.0 Array 1368371_PM_at - -
          Affymetrix HT Rat Focus Array 1368371_PM_at - -
          Affymetrix HT Rat Focus Array 1368371_at - -
          Affymetrix Rat Clariom S Assay PSR0100023633.rn.1 TC0100002934.rn.1 RTA TC
          Rat Expression Set 230 1368371_at - -
          Rat Expression Set 230 1368371_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10712530 - -
          Rat Genome U34 Set U92655_at - -
          Rat Genome U34 Set U92655_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22319178 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370502 - -
          RatRef-12 Expression BeadChip ILMN_70176 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline