Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583521_m1
          See other GLRX3 GE Assays ›
          Gene Symbol:
          Glrx3
          Gene Name:
          glutaredoxin 3
          Gene Aliases:
          PICOT, Txnl2
          Chromosome Location:
          Chr.1: 209768696 - 209799912 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583521_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Glrx3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032614.2 NP_116003.2 4 - 5 487 63
          XM_006230497.2 XP_006230559.1 4 - 5 900 63
          GenBank mRNA AF118651.1 - 4 - 5 321 63
          BC086381.1 - 4 - 5 487 7301390 63
          FQ216152.1 - 1 - 2 137 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002801.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Glrx3

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of the force of heart contraction
          negative regulation of cardiac muscle hypertrophy
          cell redox homeostasis
          oxidation-reduction process
          protein kinase C binding
          electron carrier activity
          protein disulfide oxidoreductase activity
          poly(A) RNA binding
          metal ion binding
          iron-sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P626033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27996 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P626033 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P626033 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P626033 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P432109 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P626033 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96371 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13073 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58385 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10711882 - -
          Affymetrix Rat Gene 1.0 ST Array 10711883 - -
          Affymetrix Rat Gene 1.0 ST Array 10711888 - -
          Affymetrix Rat Gene 1.0 ST Array 10711891 - -
          Affymetrix Rat Gene 1.0 ST Array 10711887 - -
          Affymetrix Rat Gene 1.0 ST Array 10711886 - -
          Affymetrix Rat Gene 1.0 ST Array 10711893 - -
          Affymetrix Rat Gene 1.0 ST Array 10711890 - -
          Affymetrix Rat Gene 1.0 ST Array 10711884 - -
          Affymetrix Rat Gene 1.0 ST Array 10711885 - -
          Affymetrix Rat Gene 1.0 ST Array 10711889 - -
          Affymetrix Rat Gene 1.0 ST Array 10711892 - -
          Affymetrix Rat Gene 1.1 ST Array 10711881 - -
          Rat Genome 230 2.0 Array 1367870_at - -
          Rat Genome 230 2.0 Array 1367870_PM_at - -
          Affymetrix HT Rat Focus Array 1367870_at - -
          Affymetrix HT Rat Focus Array 1367870_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100022051.rn.1 TC0100002801.rn.1 RTA TC
          Rat Expression Set 230 1367870_PM_at - -
          Rat Expression Set 230 1367870_at - -
          Affymetrix Rat Gene 1.0 ST Array 10711881 - -
          Rat Genome U34 Set rc_AI228347_f_at - -
          Rat Genome U34 Set rc_AI171837_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20840160 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368620 - -
          RatRef-12 Expression BeadChip ILMN_61571 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline