Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583568_m1
          See other SEC31A GE Assays ›
          Gene Symbol:
          Sec31a
          Gene Name:
          SEC31 homolog A (S. cerevisiae)
          Gene Aliases:
          Sec31l1, VAP1
          Chromosome Location:
          Chr.14: 10854692 - 10909579 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583568_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sec31a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033021.1 NP_148981.1 24 - 25 3377 60
          XM_006250676.2 XP_006250738.1 25 - 26 3461 60
          XM_006250677.1 XP_006250739.1 25 - 26 3194 60
          XM_006250678.2 XP_006250740.1 25 - 26 3416 60
          XM_006250679.2 XP_006250741.1 24 - 25 3383 60
          XM_006250680.2 XP_006250742.1 24 - 25 3377 60
          XM_006250681.2 XP_006250743.1 24 - 25 3368 60
          XM_006250682.2 XP_006250744.1 23 - 24 3344 60
          XM_006250683.2 XP_006250745.1 23 - 24 3329 60
          XM_006250684.2 XP_006250746.1 23 - 24 3284 60
          XM_006250685.2 XP_006250747.1 21 - 22 3212 60
          XM_006250686.2 XP_006250748.1 21 - 22 3167 60
          XM_006250687.2 XP_006250749.1 23 - 24 3086 60
          XM_006250688.2 XP_006250750.1 22 - 23 2993 60
          XM_006250689.2 XP_006250751.1 20 - 21 2876 60
          XM_006250690.1 XP_006250752.1 20 - 21 2610 60
          GenBank mRNA AF034582.1 - 24 - 25 3377 60
          BC085722.1 - 20 - 21 2616 7107486 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000179.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sec31a

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein transport
          vesicle-mediated transport
          response to calcium ion
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082708 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95964 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58896 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143380 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160570 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9405 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58480 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59364 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93421 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082708 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082708 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082708 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082709 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P102607 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702236079 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83411 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6534 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5233 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24025 - -
          Agilent Rat cDNA Microarray (G4105A) 702236079 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771447 - -
          Affymetrix Rat Gene 1.0 ST Array 10771448 - -
          Affymetrix Rat Gene 1.0 ST Array 10771433 - -
          Affymetrix Rat Gene 1.0 ST Array 10771439 - -
          Affymetrix Rat Gene 1.0 ST Array 10771443 - -
          Affymetrix Rat Gene 1.0 ST Array 10771442 - -
          Affymetrix Rat Gene 1.0 ST Array 10771428 - -
          Affymetrix Rat Gene 1.0 ST Array 10771453 - -
          Affymetrix Rat Gene 1.0 ST Array 10771427 - -
          Affymetrix Rat Gene 1.0 ST Array 10771429 - -
          Affymetrix Rat Gene 1.0 ST Array 10771432 - -
          Affymetrix Rat Gene 1.0 ST Array 10771430 - -
          Affymetrix Rat Gene 1.0 ST Array 10771451 - -
          Affymetrix Rat Gene 1.0 ST Array 10771435 - -
          Affymetrix Rat Gene 1.0 ST Array 10771445 - -
          Affymetrix Rat Gene 1.0 ST Array 10771437 - -
          Affymetrix Rat Gene 1.0 ST Array 10771455 - -
          Affymetrix Rat Gene 1.0 ST Array 10771446 - -
          Affymetrix Rat Gene 1.0 ST Array 10771454 - -
          Affymetrix Rat Gene 1.0 ST Array 10771434 - -
          Affymetrix Rat Gene 1.0 ST Array 10771438 - -
          Affymetrix Rat Gene 1.0 ST Array 10771449 - -
          Affymetrix Rat Gene 1.0 ST Array 10771436 - -
          Affymetrix Rat Gene 1.0 ST Array 10771444 - -
          Affymetrix Rat Gene 1.0 ST Array 10771452 - -
          Affymetrix Rat Gene 1.0 ST Array 10771440 - -
          Affymetrix Rat Gene 1.0 ST Array 10771431 - -
          Affymetrix Rat Gene 1.1 ST Array 10771426 - -
          Rat Genome 230 2.0 Array 1367753_at - -
          Rat Genome 230 2.0 Array 1367753_PM_at - -
          Affymetrix HT Rat Focus Array 1367753_at - -
          Affymetrix HT Rat Focus Array 1367753_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400001390.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001392.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001395.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001406.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001417.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001424.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001407.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001428.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001397.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001400.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001404.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001405.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001409.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001396.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001401.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001411.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001413.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001415.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001423.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001425.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001426.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001410.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001427.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001393.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001398.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001399.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001416.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001421.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001389.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001391.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001394.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001414.rn.1 TC1400000179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001422.rn.1 TC1400000179.rn.1 RTA TC
          Rat Expression Set 230 1367753_PM_at - -
          Rat Expression Set 230 1367753_at - -
          Affymetrix Rat Gene 1.0 ST Array 10771426 - -
          Rat Genome U34 Set rc_AA800517_s_at - -
          Rat Genome U34 Set AF034582_at - -
          Rat Genome U34 Set AF034582_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20847338 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370886 - -
          RatRef-12 Expression BeadChip ILMN_57540 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline