Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583657_g1
          See other HBB GE Assays ›
          Gene Symbol:
          Hbb
          Gene Name:
          hemoglobin, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 168971269 - 168972680 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583657_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hbb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033234.1 NP_150237.1 1 - 2 142 68
          GenBank mRNA BC058448.1 - 1 - 2 151 6921704 68
          FQ209544.1 - 1 - 2 147 68
          FQ209725.1 - 1 - 2 149 68
          FQ209747.1 - 1 - 2 147 68
          FQ209754.1 - 1 - 2 147 68
          FQ209760.1 - 1 - 2 147 68
          FQ209789.1 - 1 - 2 147 68
          FQ209796.1 - 1 - 2 147 68
          FQ209806.1 - 1 - 2 149 68
          FQ209822.1 - 147 68
          FQ209826.1 - 1 - 2 156 68
          FQ209841.1 - 1 - 2 149 68
          FQ209870.1 - 1 - 2 149 68
          FQ209871.1 - 1 - 2 147 68
          FQ209903.1 - 1 - 2 147 68
          FQ209929.1 - 1 - 2 147 68
          FQ209937.1 - 1 - 2 147 68
          FQ209954.1 - 1 - 2 147 68
          FQ209961.1 - 1 - 2 146 68
          FQ209989.1 - 1 - 2 147 68
          FQ209996.1 - 1 - 2 147 68
          FQ210038.1 - 1 - 2 149 68
          FQ210054.1 - 1 - 2 147 68
          FQ210058.1 - 1 - 2 147 68
          FQ210090.1 - 1 - 2 149 68
          FQ210123.1 - 1 - 2 147 68
          FQ210127.1 - 1 - 2 147 68
          FQ210137.1 - 1 - 2 147 68
          FQ210175.1 - 1 - 2 145 68
          FQ210181.1 - 1 - 2 149 68
          FQ210200.1 - 1 - 2 147 68
          FQ210216.1 - 1 - 2 146 68
          FQ210218.1 - 1 - 2 147 68
          FQ210280.1 - 1 - 2 170 68
          FQ210343.1 - 1 - 2 147 68
          FQ210348.1 - 1 - 2 147 68
          FQ210353.1 - 1 - 2 147 68
          FQ210360.1 - 1 - 2 147 68
          FQ210373.1 - 1 - 2 147 68
          FQ210415.1 - 1 - 2 147 68
          FQ210449.1 - 1 - 2 147 68
          FQ210466.1 - 1 - 2 147 68
          FQ210482.1 - 1 - 2 147 68
          FQ210486.1 - 1 - 2 149 68
          FQ210495.1 - 1 - 2 147 68
          FQ210497.1 - 1 - 2 148 68
          FQ210499.1 - 1 - 2 147 68
          FQ210511.1 - 1 - 2 147 68
          FQ210523.1 - 1 - 2 147 68
          FQ210534.1 - 1 - 2 147 68
          FQ210558.1 - 1 - 2 147 68
          FQ210602.1 - 1 - 2 147 68
          FQ210676.1 - 1 - 2 147 68
          FQ210677.1 - 1 - 2 147 68
          FQ210735.1 - 1 - 2 146 68
          FQ210771.1 - 1 - 2 147 68
          FQ210783.1 - 1 - 2 139 68
          FQ210789.1 - 1 - 2 147 68
          FQ210794.1 - 1 - 2 147 68
          FQ210798.1 - 1 - 2 147 68
          FQ210806.1 - 1 - 2 147 68
          FQ210826.1 - 1 - 2 147 68
          FQ210828.1 - 1 - 2 144 68
          FQ210829.1 - 1 - 2 147 68
          FQ210838.1 - 1 - 2 147 68
          FQ210875.1 - 1 - 2 147 68
          FQ210912.1 - 1 - 2 147 68
          FQ210937.1 - 1 - 2 147 68
          FQ210949.1 - 1 - 2 150 68
          FQ210960.1 - 1 - 2 147 68
          FQ210984.1 - 1 - 2 147 68
          FQ210985.1 - 1 - 2 147 68
          FQ210998.1 - 1 - 2 147 68
          FQ211014.1 - 1 - 2 147 68
          FQ211016.1 - 1 - 2 147 68
          FQ211034.1 - 1 - 2 147 68
          FQ211061.1 - 1 - 2 146 68
          FQ211103.1 - 1 - 2 147 68
          FQ211125.1 - 1 - 2 149 68
          FQ211133.1 - 1 - 2 144 68
          FQ211137.1 - 1 - 2 147 68
          FQ211142.1 - 1 - 2 149 68
          FQ211147.1 - 1 - 2 147 68
          FQ211154.1 - 1 - 2 147 68
          FQ211182.1 - 1 - 2 147 68
          FQ211193.1 - 1 - 2 147 68
          FQ211207.1 - 1 - 2 149 68
          FQ211241.1 - 1 - 2 147 68
          FQ211242.1 - 1 - 2 147 68
          FQ211290.1 - 1 - 2 147 68
          FQ211314.1 - 1 - 2 147 68
          FQ211331.1 - 1 - 2 147 68
          FQ211336.1 - 1 - 2 147 68
          FQ211338.1 - 1 - 2 146 68
          FQ211340.1 - 1 - 2 147 68
          FQ211443.1 - 1 - 2 147 68
          FQ211452.1 - 1 - 2 147 68
          FQ211477.1 - 1 - 2 147 68
          FQ211497.1 - 1 - 2 147 68
          FQ211548.1 - 1 - 2 147 68
          FQ211583.1 - 1 - 2 147 68
          FQ211613.1 - 1 - 2 149 68
          FQ211617.1 - 1 - 2 147 68
          FQ211626.1 - 1 - 2 147 68
          FQ211692.1 - 1 - 2 147 68
          FQ211696.1 - 1 - 2 147 68
          FQ211790.1 - 1 - 2 147 68
          FQ211806.1 - 1 - 2 147 68
          FQ211815.1 - 1 - 2 147 68
          FQ211829.1 - 1 - 2 149 68
          FQ211847.1 - 1 - 2 147 68
          FQ211858.1 - 1 - 2 147 68
          FQ211864.1 - 1 - 2 147 68
          FQ211905.1 - 1 - 2 147 68
          FQ211999.1 - 1 - 2 147 68
          FQ212127.1 - 1 - 2 147 68
          FQ212156.1 - 1 - 2 145 68
          FQ212234.1 - 1 - 2 147 68
          FQ212241.1 - 1 - 2 147 68
          FQ212258.1 - 1 - 2 146 68
          FQ212275.1 - 1 - 2 147 68
          FQ212287.1 - 1 - 2 147 68
          FQ212290.1 - 1 - 2 147 68
          FQ212303.1 - 1 - 2 147 68
          FQ212341.1 - 1 - 2 149 68
          FQ212449.1 - 1 - 2 147 68
          FQ213352.1 - 1 - 2 147 68
          FQ213466.1 - 1 - 2 147 68
          FQ213534.1 - 1 - 2 147 68
          FQ213582.1 - 626 68
          FQ213796.1 - 1 - 2 147 68
          FQ214044.1 - 1 - 2 147 68
          FQ214573.1 - 1 - 2 147 68
          FQ217014.1 - 1 - 2 149 68
          FQ217108.1 - 1 - 2 68
          FQ217151.1 - 1 - 2 147 68
          FQ217201.1 - 1 - 2 147 68
          FQ217206.1 - 1 - 2 147 68
          FQ217586.1 - 1 - 2 149 68
          FQ217619.1 - 147 68
          FQ217623.1 - 1 - 2 147 68
          FQ217653.1 - 1 - 2 147 68
          FQ217685.1 - 1 - 2 147 68
          FQ217803.1 - 1 - 2 116 68
          FQ218042.1 - 1 - 2 147 68
          FQ218100.1 - 1 - 2 147 68
          FQ218112.1 - 1 - 2 147 68
          FQ218118.1 - 1 - 2 149 68
          FQ218124.1 - 1 - 2 151 68
          FQ218145.1 - 1 - 2 147 68
          FQ218153.1 - 1 - 2 147 68
          FQ218162.1 - 1 - 2 147 68
          FQ218163.1 - 1 - 2 147 68
          FQ218171.1 - 1 - 2 147 68
          FQ218180.1 - 1 - 2 147 68
          FQ218182.1 - 1 - 2 147 68
          FQ218199.1 - 1 - 2 150 68
          FQ218229.1 - 1 - 2 150 68
          FQ218253.1 - 1 - 2 147 68
          FQ218272.1 - 1 - 2 144 68
          FQ218288.1 - 1 - 2 147 68
          FQ218461.1 - 1 - 2 167 68
          FQ218539.1 - 1 - 2 146 68
          FQ218748.1 - 1 - 2 147 68
          FQ218761.1 - 1 - 2 147 68
          FQ218763.1 - 1 - 2 147 68
          FQ218779.1 - 1 - 2 145 68
          FQ219985.1 - 1 - 2 96 68
          FQ220085.1 - 1 - 2 147 68
          FQ220253.1 - 1 - 2 96 68
          FQ221076.1 - 1 - 2 147 68
          FQ221202.1 - 1 - 2 149 68
          FQ221246.1 - 1 - 2 147 68
          FQ221520.1 - 1 - 2 147 68
          FQ221738.1 - 1 - 2 146 68
          FQ221956.1 - 1 - 2 147 68
          FQ221962.1 - 1 - 2 147 68
          FQ222032.1 - 1 - 2 147 68
          FQ222179.1 - 1 - 2 147 68
          FQ222262.1 - 1 - 2 147 68
          FQ222275.1 - 1 - 2 147 68
          FQ222291.1 - 1 - 2 147 68
          FQ222503.1 - 1 - 2 147 68
          FQ222804.1 - 1 - 2 147 68
          FQ222942.1 - 1 - 2 147 68
          FQ222960.1 - 1 - 2 147 68
          FQ223002.1 - 1 - 2 147 68
          FQ223079.1 - 1 - 2 147 68
          FQ223164.1 - 1 - 2 144 68
          FQ223490.1 - 1 - 2 147 68
          FQ223631.1 - 1 - 2 147 68
          FQ223717.1 - 1 - 2 147 68
          FQ223745.1 - 1 - 2 149 68
          FQ223805.1 - 1 - 2 147 68
          FQ223852.1 - 1 - 2 139 68
          FQ223888.1 - 1 - 2 147 68
          FQ223889.1 - 1 - 2 149 68
          FQ223995.1 - 1 - 2 120 68
          FQ224045.1 - 1 - 2 149 68
          FQ224088.1 - 1 - 2 147 68
          FQ224309.1 - 1 - 2 147 68
          FQ224343.1 - 1 - 2 147 68
          FQ224371.1 - 1 - 2 147 68
          FQ224406.1 - 1 - 2 147 68
          FQ224475.1 - 1 - 2 147 68
          FQ224682.1 - 1 - 2 147 68
          FQ224727.1 - 1 - 2 147 68
          FQ225124.1 - 1 - 2 209 68
          FQ225436.1 - 1 - 2 147 68
          FQ225684.1 - 299 68
          FQ225738.1 - 1 - 2 147 68
          FQ225754.1 - 1 - 2 146 68
          FQ225816.1 - 1 - 2 147 68
          FQ225817.1 - 1 - 2 147 68
          FQ226188.1 - 1 - 2 145 68
          FQ226198.1 - 148 68
          FQ226536.1 - 1 - 2 147 68
          FQ226572.1 - 1 - 2 147 68
          FQ226889.1 - 1 - 2 147 68
          FQ227120.1 - 1 - 2 147 68
          FQ227384.1 - 148 68
          FQ227498.1 - 1 - 2 147 68
          FQ227822.1 - 1 - 2 147 68
          FQ227875.1 - 1 - 2 147 68
          FQ228016.1 - 1 - 2 147 68
          FQ228244.1 - 1 - 2 147 68
          FQ228281.1 - 1 - 2 147 68
          FQ228294.1 - 1 - 2 147 68
          FQ228295.1 - 1 - 2 147 68
          FQ228296.1 - 1 - 2 149 68
          FQ228414.1 - 1 - 2 147 68
          FQ228616.1 - 1 - 2 147 68
          FQ228638.1 - 1 - 2 147 68
          FQ229015.1 - 1 - 2 147 68
          FQ229033.1 - 1 - 2 147 68
          FQ229489.1 - 1 - 2 147 68
          FQ230471.1 - 1 - 2 147 68
          FQ230503.1 - 1 - 2 147 68
          FQ230508.1 - 1 - 2 147 68
          FQ230541.1 - 1 - 2 147 68
          FQ230866.1 - 1 - 2 147 68
          FQ230889.1 - 1 - 2 147 68
          FQ231191.1 - 1 - 2 146 68
          FQ231199.1 - 1 - 2 147 68
          FQ231208.1 - 1 - 2 146 68
          FQ231230.1 - 1 - 2 147 68
          FQ231253.1 - 1 - 2 147 68
          FQ231284.1 - 1 - 2 149 68
          FQ231286.1 - 1 - 2 147 68
          FQ232390.1 - 415 68
          FQ232655.1 - 1 - 2 179 68
          FQ233311.1 - 1 - 2 222 68
          FQ234713.1 - 1 - 2 149 68
          FQ235254.1 - 783 68
          M17084.1 - 1 - 2 99 68
          X16417.1 - 1 - 2 142 68

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hbb

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione metabolic process
          regulation of eIF2 alpha phosphorylation by heme
          oxygen transport
          hemopoiesis
          erythrocyte development
          protein heterooligomerization
          oxygen transporter activity
          iron ion binding
          oxygen binding
          heme binding
          hemoglobin alpha binding
          hemoglobin beta binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P306307 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P028036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537049 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174610 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153469 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108342 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P306307 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028036 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P306307 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028036 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P306307 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P028036 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P306307 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103537 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724312 - -
          Affymetrix Rat Gene 1.0 ST Array 10724317 - -
          Affymetrix Rat Gene 1.0 ST Array 10724313 - -
          Affymetrix Rat Gene 1.0 ST Array 10724314 - -
          Affymetrix Rat Gene 1.0 ST Array 10724318 - -
          Affymetrix Rat Gene 1.1 ST Array 10724311 - -
          Affymetrix Rat Gene 1.1 ST Array 10724315 - -
          Rat Genome 230 2.0 Array 1367553_PM_x_at - -
          Rat Genome 230 2.0 Array 1367553_x_at - -
          Affymetrix HT Rat Focus Array 1367553_PM_x_at - -
          Affymetrix HT Rat Focus Array 1367553_x_at - -
          Affymetrix Rat Clariom S Assay PSR0100049351.rn.1 TC0100006234.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049347.rn.1 TC0100006234.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049353.rn.1 TC0100006234.rn.1 RTA TC
          Rat Expression Set 230 1367553_x_at - -
          Rat Expression Set 230 1367553_PM_x_at - -
          Affymetrix Rat Gene 1.0 ST Array 10724315 - -
          Rat Genome U34 Set M94918mRNA_f_at - -
          Rat Genome U34 Set rc_AI179576_s_at - -
          Rat Genome U34 Set rc_AI011943_at - -
          Rat Genome U34 Set rc_AI177992_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21302671 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353696 - -
          RatRef-12 Expression BeadChip ILMN_63346 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline