Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583681_m1
          See other GAL GE Assays ›
          Gene Symbol:
          Gal
          Gene Name:
          galanin/GMAP prepropeptide
          Gene Aliases:
          Galn
          Chromosome Location:
          Chr.1: 218653059 - 218657922 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583681_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gal
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033237.1 NP_150240.1 5 - 6 452 111
          GenBank mRNA J03624.1 - 5 - 6 452 111
          M18102.1 - 5 - 6 432 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007032.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Rat Estrogen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gal

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          neuropeptide signaling pathway
          nervous system development
          feeding behavior
          negative regulation of cell proliferation
          protein kinase A signaling
          cAMP-mediated signaling
          regulation of glucocorticoid metabolic process
          response to insulin
          response to immobilization stress
          response to drug
          positive regulation of apoptotic process
          response to estrogen
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of lymphocyte proliferation
          regulation of immune response
          positive regulation of cortisol secretion
          positive regulation of catagen
          positive regulation of large conductance calcium-activated potassium channel activity
          G-protein coupled receptor binding
          neuropeptide hormone activity
          type 1 galanin receptor binding
          type 2 galanin receptor binding
          type 3 galanin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P614175 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P091940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770847 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P614175 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P614175 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P091940 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P614175 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P614175 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71835 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727328 - -
          Affymetrix Rat Gene 1.0 ST Array 10727324 - -
          Affymetrix Rat Gene 1.0 ST Array 10727325 - -
          Affymetrix Rat Gene 1.0 ST Array 10727326 - -
          Affymetrix Rat Gene 1.0 ST Array 10727322 - -
          Affymetrix Rat Gene 1.0 ST Array 10727327 - -
          Affymetrix Rat Gene 1.0 ST Array 10727323 - -
          Affymetrix Rat Gene 1.1 ST Array 10727321 - -
          Rat Genome 230 2.0 Array 1387088_at - -
          Rat Genome 230 2.0 Array 1387088_PM_at - -
          Affymetrix HT Rat Focus Array 1387088_at - -
          Affymetrix HT Rat Focus Array 1387088_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100055797.rn.1 TC0100007032.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100055796.rn.1 TC0100007032.rn.1 RTA TC
          Rat Expression Set 230 1387088_at - -
          Rat Expression Set 230 1387088_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727321 - -
          Rat Genome U34 Set J03624_at - -
          Rat Genome U34 Set J03624_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22193827 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370600 - -
          RatRef-12 Expression BeadChip ILMN_49905 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline