Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583706_m1
          See other HAGH GE Assays ›
          Gene Symbol:
          Hagh
          Gene Name:
          hydroxyacyl glutathione hydrolase
          Gene Aliases:
          Glo2, RSP29
          Chromosome Location:
          Chr.10: 14215991 - 14230506 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583706_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hagh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033349.2 NP_203500.2 3 - 4 347 116
          XM_006245899.1 XP_006245961.1 3 - 4 324 116
          XM_006245900.1 XP_006245962.1 2 - 3 238 116
          GenBank mRNA BC097301.1 - 3 - 4 299 116
          CB715597.1 - 3 - 4 376 116
          FQ229806.1 - 3 - 4 416 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000225.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hagh

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          glutathione metabolic process
          glutathione biosynthetic process
          spermatogenesis
          hydroxyacylglutathione hydrolase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P156791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225108 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66636 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119033 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156791 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156791 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P156791 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12737 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14846 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64566 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10732233 - -
          Affymetrix Rat Gene 1.0 ST Array 10732236 - -
          Affymetrix Rat Gene 1.0 ST Array 10732238 - -
          Affymetrix Rat Gene 1.0 ST Array 10732229 - -
          Affymetrix Rat Gene 1.0 ST Array 10732231 - -
          Affymetrix Rat Gene 1.0 ST Array 10732230 - -
          Affymetrix Rat Gene 1.0 ST Array 10732232 - -
          Affymetrix Rat Gene 1.0 ST Array 10732237 - -
          Affymetrix Rat Gene 1.0 ST Array 10732235 - -
          Affymetrix Rat Gene 1.0 ST Array 10732234 - -
          Affymetrix Rat Gene 1.1 ST Array 10732228 - -
          Rat Genome 230 2.0 Array 1369986_PM_at - -
          Rat Genome 230 2.0 Array 1369986_at - -
          Affymetrix HT Rat Focus Array 1369986_at - -
          Affymetrix HT Rat Focus Array 1369986_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000002192.rn.1 TC1000000225.rn.1 RTA TC
          Rat Expression Set 230 1369986_at - -
          Rat Expression Set 230 1369986_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10732228 - -
          Rat Genome U34 Set rc_AI012802_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21968305 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353517 - -
          RatRef-12 Expression BeadChip ILMN_56751 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline