Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583712_m1
          See other FCGRT GE Assays ›
          Gene Symbol:
          Fcgrt
          Gene Name:
          Fc fragment of IgG, receptor, transporter, alpha
          Gene Aliases:
          FcRn
          Chromosome Location:
          Chr.1: 101086872 - 101096158 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583712_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fcgrt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033351.2 NP_203502.1 3 - 4 807 84
          XM_006229043.1 XP_006229105.1 4 - 5 916 84
          XM_008759343.1 XP_008757565.1 4 - 5 815 84
          GenBank mRNA BC061975.1 - 4 - 5 759 5623787 84
          M35495.1 - 3 - 4 809 84
          X14323.1 - 3 - 4 809 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005376.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fcgrt

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          humoral immune response
          antigen processing and presentation
          Fc-gamma receptor signaling pathway
          antigen binding
          IgG receptor activity
          IgG binding
          beta-2-microglobulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P996997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144694 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103347 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P996997 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P996997 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P996997 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P996997 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700544184 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701435672 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81267 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4143 - -
          Agilent Rat cDNA Microarray (G4105A) 700544184 - -
          Agilent Rat cDNA Microarray (G4105A) 701435672 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10721675 - -
          Affymetrix Rat Gene 1.0 ST Array 10721670 - -
          Affymetrix Rat Gene 1.0 ST Array 10721671 - -
          Affymetrix Rat Gene 1.0 ST Array 10721673 - -
          Affymetrix Rat Gene 1.0 ST Array 10721669 - -
          Affymetrix Rat Gene 1.0 ST Array 10721672 - -
          Affymetrix Rat Gene 1.0 ST Array 10721668 - -
          Affymetrix Rat Gene 1.0 ST Array 10721674 - -
          Affymetrix Rat Gene 1.1 ST Array 10721667 - -
          Rat Genome 230 2.0 Array 1379403_at - -
          Rat Genome 230 2.0 Array 1379403_PM_at - -
          Rat Genome 230 2.0 Array 1387885_PM_at - -
          Rat Genome 230 2.0 Array 1387885_at - -
          Affymetrix HT Rat Focus Array 1387885_PM_at - -
          Affymetrix HT Rat Focus Array 1387885_at - -
          Affymetrix HT Rat Focus Array 1379403_at - -
          Affymetrix HT Rat Focus Array 1379403_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100043636.rn.1 TC0100005376.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043638.rn.1 TC0100005376.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043641.rn.1 TC0100005376.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043642.rn.1 TC0100005376.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043639.rn.1 TC0100005376.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043643.rn.1 TC0100005376.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043646.rn.1 TC0100005376.rn.1 RTA TC
          Rat Expression Set 230 1387885_at - -
          Rat Expression Set 230 1387885_PM_at - -
          Rat Expression Set 230 1379403_at - -
          Rat Expression Set 230 1379403_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10721667 - -
          Rat Genome U34 Set X14323cds_g_at - -
          Rat Genome U34 Set X14323cds_at - -
          Rat Genome U34 Set rc_AI180013_at - -
          Rat Genome U34 Set rc_AI044660_at - -
          Rat Genome U34 Set rc_AI044246_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22145368 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350117 - -
          RatRef-12 Expression BeadChip ILMN_69907 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline