Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583793_m1
          See other NOS1 GE Assays ›
          Gene Symbol:
          Nos1
          Gene Name:
          nitric oxide synthase 1, neuronal
          Gene Aliases:
          bNOS
          Chromosome Location:
          Chr.12: 44215395 - 44405539 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583793_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nos1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_052799.1 NP_434686.1 6 - 7 1470 65
          XM_008769210.1 XP_008767432.1 7 - 8 2281 65
          GenBank mRNA U67309.1 - 6 - 7 1470 65
          X59949.1 - 6 - 7 1717 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001693.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Rat Hypoxia Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nos1

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          regulation of sodium ion transport
          arginine catabolic process
          nitric oxide biosynthetic process
          muscle contraction
          striated muscle contraction
          nitric oxide mediated signal transduction
          female pregnancy
          aging
          regulation of heart contraction
          negative regulation of cell proliferation
          response to heat
          response to organonitrogen compound
          response to lead ion
          response to organic cyclic compound
          response to activity
          peptidyl-cysteine S-nitrosylation
          positive regulation of guanylate cyclase activity
          response to nutrient levels
          response to lipopolysaccharide
          negative regulation of peptidyl-serine phosphorylation
          response to vitamin E
          multicellular organismal response to stress
          positive regulation of histone acetylation
          response to nicotine
          exogenous drug catabolic process
          negative regulation of apoptotic process
          negative regulation of potassium ion transport
          response to peptide hormone
          response to estrogen
          response to ethanol
          negative regulation of blood pressure
          negative regulation of heart contraction
          positive regulation of transcription, DNA-templated
          negative regulation of vasoconstriction
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of insulin secretion
          behavioral response to cocaine
          negative regulation of hydrolase activity
          negative regulation of cytosolic calcium ion concentration
          negative regulation of serotonin uptake
          negative regulation of calcium ion transport
          regulation of sensory perception of pain
          oxidation-reduction process
          cellular response to mechanical stimulus
          cellular response to growth factor stimulus
          response to nitric oxide
          cellular response to epinephrine stimulus
          positive regulation of the force of heart contraction
          positive regulation of long-term synaptic potentiation
          positive regulation of adrenergic receptor signaling pathway involved in heart process
          positive regulation of neuron death
          positive regulation of sodium ion transmembrane transport
          NADPH-hemoprotein reductase activity
          nitric-oxide synthase activity
          iron ion binding
          protein binding
          calmodulin binding
          FMN binding
          amino acid binding
          sodium channel regulator activity
          enzyme binding
          heme binding
          protein homodimerization activity
          ion channel binding
          cadmium ion binding
          flavin adenine dinucleotide binding
          NADP binding
          ATPase binding
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P473047 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133183 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P473047 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105620 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105620 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P473047 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105620 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P473047 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P473047 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42672 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85206 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27399 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10762471 - -
          Affymetrix Rat Gene 1.0 ST Array 10762463 - -
          Affymetrix Rat Gene 1.0 ST Array 10762470 - -
          Affymetrix Rat Gene 1.0 ST Array 10762461 - -
          Affymetrix Rat Gene 1.0 ST Array 10762469 - -
          Affymetrix Rat Gene 1.0 ST Array 10762462 - -
          Affymetrix Rat Gene 1.0 ST Array 10762464 - -
          Affymetrix Rat Gene 1.0 ST Array 10762466 - -
          Affymetrix Rat Gene 1.0 ST Array 10762468 - -
          Affymetrix Rat Gene 1.0 ST Array 10762458 - -
          Affymetrix Rat Gene 1.0 ST Array 10762456 - -
          Affymetrix Rat Gene 1.0 ST Array 10762467 - -
          Affymetrix Rat Gene 1.0 ST Array 10762465 - -
          Affymetrix Rat Gene 1.0 ST Array 10762457 - -
          Affymetrix Rat Gene 1.0 ST Array 10762472 - -
          Affymetrix Rat Gene 1.0 ST Array 10762459 - -
          Affymetrix Rat Gene 1.0 ST Array 10762460 - -
          Affymetrix Rat Gene 1.0 ST Array 10762473 - -
          Affymetrix Rat Gene 1.1 ST Array 10762455 - -
          Rat Genome 230 2.0 Array 1369015_PM_at - -
          Rat Genome 230 2.0 Array 1369015_at - -
          Affymetrix HT Rat Focus Array 1369015_at - -
          Affymetrix HT Rat Focus Array 1369015_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200014461.rn.1 TC1200001693.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200014462.rn.1 TC1200001693.rn.1 RTA TC
          Rat Expression Set 230 1369015_at - -
          Rat Expression Set 230 1369015_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10762455 - -
          Rat Genome U34 Set X59949cds_s_at - -
          Rat Genome U34 Set U67309_at - -
          Rat Genome U34 Set X59949cds_s_at - -
          Rat Genome U34 Set U67309_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21538034 - -
          ABI Rat Genome Survey Microarray 21823006 - -
          ABI Rat Genome Survey Microarray 22274592 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376364 - -

          Back To Top
           

          Citations (1)

          Maternal thyroid hormone: a strong repressor of neuronal nitric oxide synthase in rat embryonic neocortex.
          Authors
          Sinha RA, Pathak A, Mohan V, Bandyopadhyay S, Rastogi L, Godbole MM
          Journal
          Endocrinology - Page No 4396 (Mon Sep 01 00:00:00 EDT 2008)
          Volume (Year)
          149 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline