Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583822_m1
          See other CHRNB4 GE Assays ›
          Gene Symbol:
          Chrnb4
          Gene Name:
          cholinergic receptor, nicotinic, beta 4 (neuronal)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 59610489 - 59629073 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583822_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chrnb4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_052806.2 NP_434693.1 1 - 2 49 82
          GenBank mRNA AY574260.1 - 1 - 2 49 82
          U42976.1 - 109 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002588.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chrnb4

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          action potential
          cation transport
          smooth muscle contraction
          regulation of smooth muscle contraction
          signal transduction
          locomotory behavior
          response to nicotine
          behavioral response to nicotine
          regulation of membrane potential
          protein heterooligomerization
          synaptic transmission involved in micturition
          cation transmembrane transport
          acetylcholine-activated cation-selective channel activity
          drug binding
          acetylcholine receptor activity
          acetylcholine binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139327 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23287 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000529 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000529 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000529 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12744 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24142 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917608 - -
          Affymetrix Rat Gene 1.0 ST Array 10917616 - -
          Affymetrix Rat Gene 1.0 ST Array 10917615 - -
          Affymetrix Rat Gene 1.0 ST Array 10917613 - -
          Affymetrix Rat Gene 1.0 ST Array 10917611 - -
          Affymetrix Rat Gene 1.0 ST Array 10917610 - -
          Affymetrix Rat Gene 1.0 ST Array 10917609 - -
          Affymetrix Rat Gene 1.0 ST Array 10917612 - -
          Affymetrix Rat Gene 1.1 ST Array 10917607 - -
          Rat Genome 230 2.0 Array 1369405_a_at - -
          Rat Genome 230 2.0 Array 1398623_PM_at - -
          Rat Genome 230 2.0 Array 1398623_at - -
          Rat Genome 230 2.0 Array 1369405_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369405_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369405_a_at - -
          Affymetrix HT Rat Focus Array 1398623_PM_at - -
          Affymetrix HT Rat Focus Array 1398623_at - -
          Affymetrix Rat Clariom S Assay PSR0800020933.rn.1 TC0800002588.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020931.rn.1 TC0800002588.rn.1 RTA TC
          Rat Expression Set 230 1369405_a_at - -
          Rat Expression Set 230 1369405_PM_a_at - -
          Rat Expression Set 230 1398623_PM_at - -
          Rat Expression Set 230 1398623_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917607 - -
          Rat Genome U34 Set J05232cds_s_at - -
          Rat Genome U34 Set U42976_at - -
          Rat Genome U34 Set rc_AI236857_at - -
          Rat Genome U34 Set U42976_at - -
          Rat Genome U34 Set J05232cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21631610 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359890 - -
          RatRef-12 Expression BeadChip ILMN_52337 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline