Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583837_m1
          See other IGF1R GE Assays ›
          Gene Symbol:
          Igf1r
          Gene Name:
          insulin-like growth factor 1 receptor
          Gene Aliases:
          IGFIRC, Igfr1, JTK13
          Chromosome Location:
          Chr.1: 128924921 - 129213816 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583837_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igf1r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_052807.2 NP_434694.1 2 - 3 689 58
          XM_006229324.2 XP_006229386.1 2 - 3 689 58
          XM_008759500.1 XP_008757722.1 2 - 3 687 58
          GenBank mRNA L29232.1 - 2 - 3 689 58
          M27293.1 - 2 - 3 689 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001713.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igf1r

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          immune response
          axonogenesis
          brain development
          epidermis development
          organ morphogenesis
          negative regulation of muscle cell apoptotic process
          establishment of cell polarity
          male sex determination
          positive regulation of cell migration
          mammary gland development
          exocrine pancreas development
          neuron projection development
          positive regulation of cytokinesis
          response to vitamin E
          peptidyl-tyrosine autophosphorylation
          negative regulation of apoptotic process
          negative regulation of MAPK cascade
          positive regulation of MAPK cascade
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of DNA replication
          positive regulation of mitotic nuclear division
          regulation of JNK cascade
          protein autophosphorylation
          insulin-like growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          positive regulation of DNA metabolic process
          protein tetramerization
          protein heterooligomerization
          inactivation of MAPKK activity
          positive regulation of protein kinase B signaling
          negative regulation of protein kinase B signaling
          prostate gland epithelium morphogenesis
          positive regulation of steroid hormone biosynthetic process
          G-protein alpha-subunit binding
          protein tyrosine kinase activity
          insulin-like growth factor-activated receptor activity
          insulin receptor binding
          insulin-like growth factor binding
          ATP binding
          insulin-like growth factor I binding
          identical protein binding
          phosphatidylinositol 3-kinase binding
          insulin binding
          insulin receptor substrate binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0100013779.rn.1 TC0100001713.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21679085 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline