Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583920_m1
          See other REG3B GE Assays ›
          Gene Symbol:
          Reg3b
          Gene Name:
          regenerating islet-derived 3 beta
          Gene Aliases:
          Pap, Pap1
          Chromosome Location:
          Chr.4: 109467272 - 109470510 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583920_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Reg3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053289.1 NP_445741.1 5 - 6 513 77
          GenBank mRNA FQ232415.1 - 5 - 6 513 77
          FQ233015.1 - 5 - 6 513 77
          M55149.1 - 5 - 6 515 77
          M98049.1 - 4 - 5 463 77
          S43715.1 - 5 - 6 513 77
          S77413.1 - 5 - 6 511 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001196.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Reg3b

          Gene Ontology Categories:

          Function(s) Process(es)

          acute-phase response
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P271658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142200 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P271658 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P271658 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P271658 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P271658 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104103 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10856477 - -
          Affymetrix Rat Gene 1.0 ST Array 10856479 - -
          Affymetrix Rat Gene 1.0 ST Array 10856480 - -
          Affymetrix Rat Gene 1.0 ST Array 10856478 - -
          Affymetrix Rat Gene 1.0 ST Array 10856475 - -
          Affymetrix Rat Gene 1.0 ST Array 10856476 - -
          Affymetrix Rat Gene 1.1 ST Array 10856474 - -
          Rat Genome 230 2.0 Array 1368238_at - -
          Rat Genome 230 2.0 Array 1368238_PM_at - -
          Affymetrix HT Rat Focus Array 1368238_at - -
          Affymetrix HT Rat Focus Array 1368238_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400008335.rn.1 TC0400001196.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400008337.rn.1 TC0400001196.rn.1 RTA TC
          Rat Expression Set 230 1368238_PM_at - -
          Rat Expression Set 230 1368238_at - -
          Affymetrix Rat Gene 1.0 ST Array 10856474 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20906301 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357508 - -
          RatRef-12 Expression BeadChip ILMN_60043 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline