Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583966_m1
          See other GLRB GE Assays ›
          Gene Symbol:
          Glrb
          Gene Name:
          glycine receptor, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 179768040 - 179842612 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583966_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Glrb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053296.1 NP_445748.1 7 - 8 907 60
          XM_006232506.2 XP_006232568.1 8 - 9 1053 60
          GenBank mRNA AJ310839.1 - 7 - 8 907 60
          FQ212388.1 - 8 - 9 1058 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003922.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Glrb

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          startle response
          ion transport
          neuropeptide signaling pathway
          chemical synaptic transmission
          acrosome reaction
          nervous system development
          visual perception
          adult walking behavior
          ion transmembrane transport
          regulation of membrane potential
          neuromuscular process
          protein heterooligomerization
          synaptic transmission, glycinergic
          righting reflex
          neurotransmitter-gated ion channel clustering
          gamma-aminobutyric acid receptor clustering
          chloride transmembrane transport
          protein binding
          drug binding
          glycine binding
          extracellular-glycine-gated ion channel activity
          extracellular-glycine-gated chloride channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P162099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201038 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96274 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37836 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136247 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161917 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86085 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10620 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P162099 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P162099 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P162099 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P162099 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74416 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74309 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31552 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39405 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73766 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9616 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823884 - -
          Affymetrix Rat Gene 1.0 ST Array 10823879 - -
          Affymetrix Rat Gene 1.0 ST Array 10823876 - -
          Affymetrix Rat Gene 1.0 ST Array 10823882 - -
          Affymetrix Rat Gene 1.0 ST Array 10823878 - -
          Affymetrix Rat Gene 1.0 ST Array 10823880 - -
          Affymetrix Rat Gene 1.0 ST Array 10823881 - -
          Affymetrix Rat Gene 1.0 ST Array 10823877 - -
          Affymetrix Rat Gene 1.0 ST Array 10823883 - -
          Affymetrix Rat Gene 1.1 ST Array 10823875 - -
          Rat Genome 230 2.0 Array 1370146_at - -
          Rat Genome 230 2.0 Array 1370146_PM_at - -
          Affymetrix HT Rat Focus Array 1370146_at - -
          Affymetrix HT Rat Focus Array 1370146_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200026337.rn.1 TC0200003922.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026340.rn.1 TC0200003922.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026336.rn.1 TC0200003922.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026338.rn.1 TC0200003922.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026334.rn.1 TC0200003922.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026339.rn.1 TC0200003922.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026332.rn.1 TC0200003922.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026335.rn.1 TC0200003922.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026333.rn.1 TC0200003922.rn.1 RTA TC
          Rat Expression Set 230 1370146_at - -
          Rat Expression Set 230 1370146_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823875 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20693868 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367994 - -
          RatRef-12 Expression BeadChip ILMN_60765 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline