Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584271_m1
          See other ACOX3 GE Assays ›
          Gene Symbol:
          Acox3
          Gene Name:
          acyl-CoA oxidase 3, pristanoyl
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 80313202 - 80358130 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584271_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acox3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053339.1 NP_445791.1 17 - 18 2022 81
          XM_008770372.1 XP_008768594.1 17 - 18 4782 81
          XM_008770373.1 XP_008768595.1 17 - 18 4825 81
          XM_008770374.1 XP_008768596.1 16 - 17 2049 81
          GenBank mRNA X95188.1 - 17 - 18 2022 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002068.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acox3

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid beta-oxidation
          fatty acid beta-oxidation using acyl-CoA oxidase
          acyl-CoA dehydrogenase activity
          acyl-CoA oxidase activity
          receptor binding
          fatty acid binding
          pristanoyl-CoA oxidase activity
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P493946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148948 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P493946 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P493946 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P493946 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P493946 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700330996 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42752 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93323 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31272 - -
          Agilent Rat cDNA Microarray (G4105A) 701478506 - -
          Agilent Rat cDNA Microarray (G4105A) 700330996 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777495 - -
          Affymetrix Rat Gene 1.0 ST Array 10777509 - -
          Affymetrix Rat Gene 1.0 ST Array 10777498 - -
          Affymetrix Rat Gene 1.0 ST Array 10777500 - -
          Affymetrix Rat Gene 1.0 ST Array 10777497 - -
          Affymetrix Rat Gene 1.0 ST Array 10777499 - -
          Affymetrix Rat Gene 1.0 ST Array 10777510 - -
          Affymetrix Rat Gene 1.0 ST Array 10777502 - -
          Affymetrix Rat Gene 1.0 ST Array 10777507 - -
          Affymetrix Rat Gene 1.0 ST Array 10777504 - -
          Affymetrix Rat Gene 1.0 ST Array 10777506 - -
          Affymetrix Rat Gene 1.0 ST Array 10777501 - -
          Affymetrix Rat Gene 1.0 ST Array 10777503 - -
          Affymetrix Rat Gene 1.0 ST Array 10777493 - -
          Affymetrix Rat Gene 1.0 ST Array 10777505 - -
          Affymetrix Rat Gene 1.0 ST Array 10777496 - -
          Affymetrix Rat Gene 1.0 ST Array 10777508 - -
          Affymetrix Rat Gene 1.1 ST Array 10777492 - -
          Rat Genome 230 2.0 Array 1377887_at - -
          Rat Genome 230 2.0 Array 1369734_PM_at - -
          Rat Genome 230 2.0 Array 1369734_at - -
          Rat Genome 230 2.0 Array 1377887_PM_at - -
          Affymetrix HT Rat Focus Array 1369734_PM_at - -
          Affymetrix HT Rat Focus Array 1369734_at - -
          Affymetrix Rat Clariom S Assay PSR1400014701.rn.1 TC1400002068.rn.1 RTA TC
          Rat Expression Set 230 1369734_at - -
          Rat Expression Set 230 1369734_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777492 - -
          Rat Genome U34 Set X95188_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21732053 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350613 - -
          RatRef-12 Expression BeadChip ILMN_62541 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline