Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584280_m1
          See other OGFR GE Assays ›
          Gene Symbol:
          Ogfr
          Gene Name:
          opioid growth factor receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 176093662 - 176099949 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584280_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ogfr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053340.2 NP_445792.2 4 - 5 404 74
          GenBank mRNA BC101898.1 - 4 - 5 385 7932932 74
          EV778659.1 - 4 - 5 404 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002570.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ogfr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of growth
          receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871049 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152662 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152662 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152662 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P233612 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19781 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842846 - -
          Affymetrix Rat Gene 1.0 ST Array 10842843 - -
          Affymetrix Rat Gene 1.0 ST Array 10842847 - -
          Affymetrix Rat Gene 1.0 ST Array 10842845 - -
          Affymetrix Rat Gene 1.0 ST Array 10842844 - -
          Affymetrix Rat Gene 1.0 ST Array 10842842 - -
          Affymetrix Rat Gene 1.0 ST Array 10842848 - -
          Affymetrix Rat Gene 1.0 ST Array 10842840 - -
          Affymetrix Rat Gene 1.0 ST Array 10842841 - -
          Affymetrix Rat Gene 1.1 ST Array 10842839 - -
          Rat Genome 230 2.0 Array 1386986_PM_at - -
          Rat Genome 230 2.0 Array 1386986_at - -
          Affymetrix HT Rat Focus Array 1386986_at - -
          Affymetrix HT Rat Focus Array 1386986_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300018920.rn.1 TC0300002570.rn.1 RTA TC
          Rat Expression Set 230 1386986_PM_at - -
          Rat Expression Set 230 1386986_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842839 - -
          Rat Genome U34 Set rc_AI044795_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21735288 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline