Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584379_m1
          See other IKBKB GE Assays ›
          Gene Symbol:
          Ikbkb
          Gene Name:
          inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
          Gene Aliases:
          AIM-1, IKK2
          Chromosome Location:
          Chr.16: 74177233 - 74230809 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584379_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ikbkb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053355.2 NP_445807.2 7 - 8 680 84
          GenBank mRNA AF115282.1 - 6 - 7 594 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000871.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ikbkb

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          B cell homeostasis
          skeletal muscle contraction
          protein phosphorylation
          I-kappaB phosphorylation
          positive regulation of cell proliferation
          response to toxic substance
          positive regulation of sodium ion transport
          neuron projection development
          response to drug
          serine phosphorylation of STAT protein
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of NF-kappaB transcription factor activity
          response to fatty acid
          cellular response to tumor necrosis factor
          positive regulation of neuron death
          positive regulation of cation channel activity
          protein kinase activity
          protein binding
          ATP binding
          IkappaB kinase activity
          protein kinase binding
          protein homodimerization activity
          protein heterodimerization activity
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136181 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18410 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12782 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12782 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12782 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12782 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700314629 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74821 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94686 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55443 - -
          Agilent Rat cDNA Microarray (G4105A) 700314629 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10789105 - -
          Affymetrix Rat Gene 1.0 ST Array 10789088 - -
          Affymetrix Rat Gene 1.0 ST Array 10789104 - -
          Affymetrix Rat Gene 1.0 ST Array 10789102 - -
          Affymetrix Rat Gene 1.0 ST Array 10789107 - -
          Affymetrix Rat Gene 1.0 ST Array 10789097 - -
          Affymetrix Rat Gene 1.0 ST Array 10789090 - -
          Affymetrix Rat Gene 1.0 ST Array 10789101 - -
          Affymetrix Rat Gene 1.0 ST Array 10789089 - -
          Affymetrix Rat Gene 1.0 ST Array 10789096 - -
          Affymetrix Rat Gene 1.0 ST Array 10789109 - -
          Affymetrix Rat Gene 1.0 ST Array 10789091 - -
          Affymetrix Rat Gene 1.0 ST Array 10789093 - -
          Affymetrix Rat Gene 1.0 ST Array 10789106 - -
          Affymetrix Rat Gene 1.0 ST Array 10789095 - -
          Affymetrix Rat Gene 1.0 ST Array 10789103 - -
          Affymetrix Rat Gene 1.0 ST Array 10789092 - -
          Affymetrix Rat Gene 1.0 ST Array 10789099 - -
          Affymetrix Rat Gene 1.0 ST Array 10789108 - -
          Affymetrix Rat Gene 1.0 ST Array 10789098 - -
          Affymetrix Rat Gene 1.0 ST Array 10789100 - -
          Affymetrix Rat Gene 1.1 ST Array 10789087 - -
          Rat Genome 230 2.0 Array 1397547_PM_at - -
          Rat Genome 230 2.0 Array 1368424_at - -
          Rat Genome 230 2.0 Array 1397547_at - -
          Rat Genome 230 2.0 Array 1368424_PM_at - -
          Affymetrix HT Rat Focus Array 1368424_PM_at - -
          Affymetrix HT Rat Focus Array 1368424_at - -
          Affymetrix HT Rat Focus Array 1397547_at - -
          Affymetrix HT Rat Focus Array 1397547_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600006680.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006682.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006687.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006684.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006692.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006693.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006695.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006676.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006688.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006677.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006691.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006678.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006689.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006679.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006683.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006685.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006686.rn.1 TC1600000871.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006696.rn.1 TC1600000871.rn.1 RTA TC
          Rat Expression Set 230 1368424_PM_at - -
          Rat Expression Set 230 1368424_at - -
          Rat Expression Set 230 1397547_PM_at - -
          Rat Expression Set 230 1397547_at - -
          Affymetrix Rat Gene 1.0 ST Array 10789087 - -
          Rat Genome U34 Set rc_AI137901_at - -
          Rat Genome U34 Set rc_AI179234_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21071525 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357714 - -
          RatRef-12 Expression BeadChip ILMN_55426 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline