Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584575_m1
          See other TLE3 GE Assays ›
          Gene Symbol:
          Tle3
          Gene Name:
          transducin-like enhancer of split 3
          Gene Aliases:
          Esp3
          Chromosome Location:
          Chr.8: 66296508 - 66342186 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584575_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tle3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053400.1 NP_445852.1 8 - 9 1338 63
          XM_006243211.2 XP_006243273.1 9 - 10 1561 63
          XM_006243212.2 XP_006243274.1 8 - 9 1734 63
          XM_006243213.2 XP_006243275.1 8 - 9 1736 63
          XM_006243214.2 XP_006243276.1 8 - 9 1737 63
          XM_006243215.2 XP_006243277.1 8 - 9 1736 63
          XM_006243216.2 XP_006243278.1 8 - 9 1706 63
          XM_006243217.2 XP_006243279.1 8 - 9 1703 63
          XM_006243218.2 XP_006243280.1 8 - 9 1705 63
          XM_006243219.2 XP_006243281.1 8 - 9 1706 63
          XM_006243220.2 XP_006243282.1 8 - 9 1703 63
          XM_006243221.2 XP_006243283.1 8 - 9 1703 63
          XM_008766347.1 XP_008764569.1 8 - 9 1736 63
          GenBank mRNA AF186092.1 - 8 - 9 1338 63
          BC097419.1 - 8 - 9 819 7454629 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000872.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Wnt Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tle3

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          Wnt signaling pathway
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P550344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7496 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137111 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33269 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P550344 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P550344 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P550344 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P550344 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701647295 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2192 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26809 - -
          Agilent Rat cDNA Microarray (G4105A) 701647295 - -
          Agilent Rat cDNA Microarray (G4105A) 702168491 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10910652 - -
          Affymetrix Rat Gene 1.0 ST Array 10910648 - -
          Affymetrix Rat Gene 1.0 ST Array 10910650 - -
          Affymetrix Rat Gene 1.0 ST Array 10910646 - -
          Affymetrix Rat Gene 1.0 ST Array 10910656 - -
          Affymetrix Rat Gene 1.0 ST Array 10910647 - -
          Affymetrix Rat Gene 1.0 ST Array 10910644 - -
          Affymetrix Rat Gene 1.0 ST Array 10910642 - -
          Affymetrix Rat Gene 1.0 ST Array 10910661 - -
          Affymetrix Rat Gene 1.0 ST Array 10910659 - -
          Affymetrix Rat Gene 1.0 ST Array 10910663 - -
          Affymetrix Rat Gene 1.0 ST Array 10910657 - -
          Affymetrix Rat Gene 1.0 ST Array 10910645 - -
          Affymetrix Rat Gene 1.0 ST Array 10910653 - -
          Affymetrix Rat Gene 1.0 ST Array 10910651 - -
          Affymetrix Rat Gene 1.0 ST Array 10910660 - -
          Affymetrix Rat Gene 1.0 ST Array 10910643 - -
          Affymetrix Rat Gene 1.0 ST Array 10910662 - -
          Affymetrix Rat Gene 1.0 ST Array 10910658 - -
          Affymetrix Rat Gene 1.0 ST Array 10910654 - -
          Affymetrix Rat Gene 1.0 ST Array 10910655 - -
          Affymetrix Rat Gene 1.1 ST Array 10910641 - -
          Rat Genome 230 2.0 Array 1387169_at - -
          Rat Genome 230 2.0 Array 1387169_PM_at - -
          Affymetrix HT Rat Focus Array 1387169_at - -
          Affymetrix HT Rat Focus Array 1387169_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800006138.rn.1 TC0800000872.rn.1 RTA TC
          Rat Expression Set 230 1387169_PM_at - -
          Rat Expression Set 230 1387169_at - -
          Affymetrix Rat Gene 1.0 ST Array 10910641 - -
          Rat Genome U34 Set rc_AA963862_at - -
          Rat Genome U34 Set rc_AA943427_at - -
          Rat Genome U34 Set rc_AI043969_at - -
          Rat Genome U34 Set rc_AI171940_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21869990 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364558 - -
          RatRef-12 Expression BeadChip ILMN_63807 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline