Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584703_m1
          See other GAB2 GE Assays ›
          Gene Symbol:
          Gab2
          Gene Name:
          GRB2-associated binding protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 162083038 - 162282707 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584703_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gab2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053417.1 NP_445869.1 2 - 3 376 74
          XM_008759806.1 XP_008758028.1 1 - 2 2008 74
          GenBank mRNA AF230367.2 - 2 - 3 376 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002070.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gab2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transmembrane receptor protein tyrosine kinase signaling pathway
          integrin-mediated signaling pathway
          positive regulation of cell proliferation
          cell migration
          osteoclast differentiation
          intracellular signal transduction
          positive regulation of mast cell degranulation
          phosphatidylinositol-mediated signaling
          transmembrane receptor protein tyrosine kinase adaptor activity
          phosphatidylinositol-3,4,5-trisphosphate binding
          protein domain specific binding
          phosphatidylinositol-3,4-bisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P144964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167977 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54937 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109468 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13572 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67679 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108583 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3828 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162366 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71455 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121292 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119117 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22554 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137130 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144964 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144964 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P144964 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P529077 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82610 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99560 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70597 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59473 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84337 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22478 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95408 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61736 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85685 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98969 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10708755 - -
          Affymetrix Rat Gene 1.0 ST Array 10708750 - -
          Affymetrix Rat Gene 1.0 ST Array 10708746 - -
          Affymetrix Rat Gene 1.0 ST Array 10708751 - -
          Affymetrix Rat Gene 1.0 ST Array 10708748 - -
          Affymetrix Rat Gene 1.0 ST Array 10708753 - -
          Affymetrix Rat Gene 1.0 ST Array 10708754 - -
          Affymetrix Rat Gene 1.0 ST Array 10708749 - -
          Affymetrix Rat Gene 1.0 ST Array 10708747 - -
          Affymetrix Rat Gene 1.0 ST Array 10708752 - -
          Affymetrix Rat Gene 1.1 ST Array 10708745 - -
          Rat Genome 230 2.0 Array 1369458_PM_at - -
          Rat Genome 230 2.0 Array 1369458_at - -
          Affymetrix HT Rat Focus Array 1369458_at - -
          Affymetrix HT Rat Focus Array 1369458_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100015873.rn.1 TC0100002070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100015872.rn.1 TC0100002070.rn.1 RTA TC
          Rat Expression Set 230 1369458_PM_at - -
          Rat Expression Set 230 1369458_at - -
          Affymetrix Rat Gene 1.0 ST Array 10708745 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21665263 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353793 - -
          RatRef-12 Expression BeadChip ILMN_61395 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline