Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584932_m1
          See other RGS2 GE Assays ›
          Gene Symbol:
          Rgs2
          Gene Name:
          regulator of G-protein signaling 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 60846458 - 60849061 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584932_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgs2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053453.2 NP_445905.1 1 - 2 103 70
          GenBank mRNA AF279918.1 - 116 70
          AF321837.1 - 1 - 2 103 70
          AJ318489.1 - 1 - 2 103 70
          AY043246.1 - 127 70
          BC061969.1 - 124 5621945 70
          FQ224353.1 - 134 70
          FQ228949.1 - 134 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001553.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rgs2

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translation
          spermatogenesis
          biological_process
          negative regulation of phospholipase activity
          negative regulation of cardiac muscle hypertrophy
          positive regulation of microtubule polymerization
          termination of G-protein coupled receptor signaling pathway
          negative regulation of MAP kinase activity
          positive regulation of GTPase activity
          negative regulation of cAMP-mediated signaling
          negative regulation of G-protein coupled receptor protein signaling pathway
          brown fat cell differentiation
          relaxation of cardiac muscle
          relaxation of vascular smooth muscle
          positive regulation of cardiac muscle contraction
          regulation of adrenergic receptor signaling pathway
          molecular_function
          GTPase activator activity
          beta-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P002156 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002156 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002156 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14131 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P002156 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14131 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14131 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701744546 - -
          Agilent Rat cDNA Microarray (G4105A) 701744546 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10768334 - -
          Affymetrix Rat Gene 1.0 ST Array 10768335 - -
          Affymetrix Rat Gene 1.0 ST Array 10768337 - -
          Affymetrix Rat Gene 1.0 ST Array 10768333 - -
          Affymetrix Rat Gene 1.0 ST Array 10768336 - -
          Affymetrix Rat Gene 1.0 ST Array 10768338 - -
          Affymetrix Rat Gene 1.1 ST Array 10768332 - -
          Rat Genome 230 2.0 Array 1387074_at - -
          Rat Genome 230 2.0 Array 1368144_PM_at - -
          Rat Genome 230 2.0 Array 1387074_PM_at - -
          Rat Genome 230 2.0 Array 1368144_at - -
          Affymetrix HT Rat Focus Array 1387074_PM_at - -
          Affymetrix HT Rat Focus Array 1387074_at - -
          Affymetrix HT Rat Focus Array 1368144_at - -
          Affymetrix HT Rat Focus Array 1368144_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300010691.rn.1 TC1300001553.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010684.rn.1 TC1300001553.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010686.rn.1 TC1300001553.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010689.rn.1 TC1300001553.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010693.rn.1 TC1300001553.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010694.rn.1 TC1300001553.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010687.rn.1 TC1300001553.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010685.rn.1 TC1300001553.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010688.rn.1 TC1300001553.rn.1 RTA TC
          Rat Expression Set 230 1368144_PM_at - -
          Rat Expression Set 230 1387074_at - -
          Rat Expression Set 230 1387074_PM_at - -
          Rat Expression Set 230 1368144_at - -
          Affymetrix Rat Gene 1.0 ST Array 10768332 - -
          Rat Genome U34 Set rc_AA858950_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20830772 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362451 - -
          RatRef-12 Expression BeadChip ILMN_48055 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0