Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585001_m1
          See other SEMA6B GE Assays ›
          Gene Symbol:
          Sema6b
          Gene Name:
          sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 10934432 - 10951252 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585001_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sema6b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053471.2 NP_445923.1 8 - 9 986 79
          XM_006244348.2 XP_006244410.1 9 - 10 1455 79
          XM_006244349.2 XP_006244411.1 8 - 9 808 79
          GenBank mRNA AB000776.1 - 8 - 9 986 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000115.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sema6b

          Panther Classification:

          Molecular Function -

          membrane-bound signaling molecule intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          cell differentiation
          receptor activity
          semaphorin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P133188 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133188 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133188 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P133188 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P464489 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71798 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79888 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10931029 - -
          Affymetrix Rat Gene 1.0 ST Array 10931020 - -
          Affymetrix Rat Gene 1.0 ST Array 10931025 - -
          Affymetrix Rat Gene 1.0 ST Array 10931023 - -
          Affymetrix Rat Gene 1.0 ST Array 10931026 - -
          Affymetrix Rat Gene 1.0 ST Array 10931022 - -
          Affymetrix Rat Gene 1.0 ST Array 10931019 - -
          Affymetrix Rat Gene 1.0 ST Array 10931028 - -
          Affymetrix Rat Gene 1.0 ST Array 10931032 - -
          Affymetrix Rat Gene 1.0 ST Array 10931027 - -
          Affymetrix Rat Gene 1.0 ST Array 10931024 - -
          Affymetrix Rat Gene 1.0 ST Array 10931018 - -
          Affymetrix Rat Gene 1.0 ST Array 10931021 - -
          Affymetrix Rat Gene 1.0 ST Array 10931030 - -
          Affymetrix Rat Gene 1.0 ST Array 10931033 - -
          Affymetrix Rat Gene 1.0 ST Array 10931031 - -
          Affymetrix Rat Gene 1.1 ST Array 10931017 - -
          Rat Genome 230 2.0 Array 1387322_PM_at - -
          Rat Genome 230 2.0 Array 1387322_at - -
          Affymetrix HT Rat Focus Array 1387322_PM_at - -
          Affymetrix HT Rat Focus Array 1387322_at - -
          Affymetrix Rat Clariom S Assay PSR0900000878.rn.1 TC0900000115.rn.1 RTA TC
          Rat Expression Set 230 1387322_PM_at - -
          Rat Expression Set 230 1387322_at - -
          Affymetrix Rat Gene 1.0 ST Array 10931017 - -
          Rat Genome U34 Set AB000776_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21972816 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650011 - -
          RatRef-12 Expression BeadChip ILMN_62550 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0