Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585042_m1
          See other SYNJ1 GE Assays ›
          Gene Symbol:
          Synj1
          Gene Name:
          synaptojanin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 31105784 - 31181573 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585042_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Synj1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053476.2 NP_445928.2 28 - 29 3670 109
          GenBank mRNA AJ006855.1 - 20 - 21 2673 109
          U45479.2 - 28 - 29 3670 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001294.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Synj1

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter transport
          brain development
          learning
          positive regulation of gliogenesis
          synaptic vesicle priming
          synaptic vesicle uncoating
          response to retinoic acid
          response to cytokine
          phosphatidylinositol metabolic process
          inositol phosphate dephosphorylation
          phosphatidylinositol dephosphorylation
          positive regulation of receptor-mediated endocytosis
          synaptic vesicle endocytosis
          synaptic vesicle transport
          nucleotide binding
          nucleic acid binding
          RNA binding
          phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity
          protein binding
          protein C-terminus binding
          SH3 domain binding
          protein complex binding
          phosphatidylinositol phosphate 5-phosphatase activity
          phosphoric ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P089886 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P153234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5807 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56212 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75805 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120995 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98070 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089886 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153234 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P153234 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089886 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P089886 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P153234 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12816 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P203421 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P395044 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701878990 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76720 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42922 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7673 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97604 - -
          Agilent Rat cDNA Microarray (G4105A) 701878990 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10753106 - -
          Affymetrix Rat Gene 1.0 ST Array 10753092 - -
          Affymetrix Rat Gene 1.0 ST Array 10753094 - -
          Affymetrix Rat Gene 1.0 ST Array 10753087 - -
          Affymetrix Rat Gene 1.0 ST Array 10753077 - -
          Affymetrix Rat Gene 1.0 ST Array 10753079 - -
          Affymetrix Rat Gene 1.0 ST Array 10753093 - -
          Affymetrix Rat Gene 1.0 ST Array 10753084 - -
          Affymetrix Rat Gene 1.0 ST Array 10753097 - -
          Affymetrix Rat Gene 1.0 ST Array 10753081 - -
          Affymetrix Rat Gene 1.0 ST Array 10753104 - -
          Affymetrix Rat Gene 1.0 ST Array 10753105 - -
          Affymetrix Rat Gene 1.0 ST Array 10753098 - -
          Affymetrix Rat Gene 1.0 ST Array 10753089 - -
          Affymetrix Rat Gene 1.0 ST Array 10753085 - -
          Affymetrix Rat Gene 1.0 ST Array 10753086 - -
          Affymetrix Rat Gene 1.0 ST Array 10753078 - -
          Affymetrix Rat Gene 1.0 ST Array 10753076 - -
          Affymetrix Rat Gene 1.0 ST Array 10753103 - -
          Affymetrix Rat Gene 1.0 ST Array 10753090 - -
          Affymetrix Rat Gene 1.0 ST Array 10753099 - -
          Affymetrix Rat Gene 1.0 ST Array 10753080 - -
          Affymetrix Rat Gene 1.0 ST Array 10753096 - -
          Affymetrix Rat Gene 1.0 ST Array 10753083 - -
          Affymetrix Rat Gene 1.0 ST Array 10753095 - -
          Affymetrix Rat Gene 1.0 ST Array 10753088 - -
          Affymetrix Rat Gene 1.0 ST Array 10753100 - -
          Affymetrix Rat Gene 1.0 ST Array 10753101 - -
          Affymetrix Rat Gene 1.0 ST Array 10753091 - -
          Affymetrix Rat Gene 1.0 ST Array 10753102 - -
          Affymetrix Rat Gene 1.1 ST Array 10753075 - -
          Rat Genome 230 2.0 Array 1370070_PM_at - -
          Rat Genome 230 2.0 Array 1388586_PM_at - -
          Rat Genome 230 2.0 Array 1388586_at - -
          Rat Genome 230 2.0 Array 1370070_at - -
          Affymetrix HT Rat Focus Array 1370070_PM_at - -
          Affymetrix HT Rat Focus Array 1388586_PM_at - -
          Affymetrix HT Rat Focus Array 1388586_at - -
          Affymetrix HT Rat Focus Array 1370070_at - -
          Affymetrix Rat Clariom S Assay PSR1100007566.rn.1 TC1100001294.rn.1 RTA TC
          Rat Expression Set 230 1388586_at - -
          Rat Expression Set 230 1370070_PM_at - -
          Rat Expression Set 230 1388586_PM_at - -
          Rat Expression Set 230 1370070_at - -
          Affymetrix Rat Gene 1.0 ST Array 10753075 - -
          Rat Genome U34 Set AJ006855_s_at - -
          Rat Genome U34 Set U45479_at - -
          Rat Genome U34 Set AJ006855_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20874049 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355054 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline