Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585063_m1
          See other PLCB2 GE Assays ›
          Gene Symbol:
          Plcb2
          Gene Name:
          phospholipase C, beta 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 110497760 - 110517261 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585063_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plcb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053478.1 NP_445930.1 16 - 17 1804 68
          GenBank mRNA AJ011035.1 - 16 - 17 1804 68
          DQ120505.1 - 16 - 17 1730 68
          DQ120506.1 - 16 - 17 1730 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003963.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plcb2

          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          metabolic process
          phospholipid catabolic process
          intracellular signal transduction
          sensory perception of bitter taste
          phosphatidylinositol phospholipase C activity
          phospholipase C activity
          signal transducer activity
          calcium ion binding
          phospholipid binding
          G-protein beta/gamma-subunit complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30937 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116237 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097704 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097704 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097704 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P165543 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65222 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73999 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10848478 - -
          Affymetrix Rat Gene 1.0 ST Array 10848477 - -
          Affymetrix Rat Gene 1.0 ST Array 10848456 - -
          Affymetrix Rat Gene 1.0 ST Array 10848479 - -
          Affymetrix Rat Gene 1.0 ST Array 10848472 - -
          Affymetrix Rat Gene 1.0 ST Array 10848460 - -
          Affymetrix Rat Gene 1.0 ST Array 10848466 - -
          Affymetrix Rat Gene 1.0 ST Array 10848454 - -
          Affymetrix Rat Gene 1.0 ST Array 10848483 - -
          Affymetrix Rat Gene 1.0 ST Array 10848461 - -
          Affymetrix Rat Gene 1.0 ST Array 10848452 - -
          Affymetrix Rat Gene 1.0 ST Array 10848462 - -
          Affymetrix Rat Gene 1.0 ST Array 10848481 - -
          Affymetrix Rat Gene 1.0 ST Array 10848468 - -
          Affymetrix Rat Gene 1.0 ST Array 10848476 - -
          Affymetrix Rat Gene 1.0 ST Array 10848470 - -
          Affymetrix Rat Gene 1.0 ST Array 10848455 - -
          Affymetrix Rat Gene 1.0 ST Array 10848482 - -
          Affymetrix Rat Gene 1.0 ST Array 10848474 - -
          Affymetrix Rat Gene 1.0 ST Array 10848458 - -
          Affymetrix Rat Gene 1.0 ST Array 10848453 - -
          Affymetrix Rat Gene 1.0 ST Array 10848459 - -
          Affymetrix Rat Gene 1.0 ST Array 10848471 - -
          Affymetrix Rat Gene 1.0 ST Array 10848480 - -
          Affymetrix Rat Gene 1.0 ST Array 10848485 - -
          Affymetrix Rat Gene 1.0 ST Array 10848465 - -
          Affymetrix Rat Gene 1.0 ST Array 10848473 - -
          Affymetrix Rat Gene 1.0 ST Array 10848475 - -
          Affymetrix Rat Gene 1.0 ST Array 10848469 - -
          Affymetrix Rat Gene 1.0 ST Array 10848467 - -
          Affymetrix Rat Gene 1.0 ST Array 10848464 - -
          Affymetrix Rat Gene 1.0 ST Array 10848463 - -
          Affymetrix Rat Gene 1.0 ST Array 10848484 - -
          Affymetrix Rat Gene 1.1 ST Array 10848450 - -
          Rat Genome 230 2.0 Array 1387717_PM_at - -
          Rat Genome 230 2.0 Array 1387717_at - -
          Affymetrix HT Rat Focus Array 1387717_PM_at - -
          Affymetrix HT Rat Focus Array 1387717_at - -
          Affymetrix Rat Clariom S Assay PSR0300030005.rn.1 TC0300003963.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300030006.rn.1 TC0300003963.rn.1 RTA TC
          Rat Expression Set 230 1387717_PM_at - -
          Rat Expression Set 230 1387717_at - -
          Affymetrix Rat Gene 1.0 ST Array 10848450 - -
          Rat Genome U34 Set AJ011035_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20965293 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369041 - -
          RatRef-12 Expression BeadChip ILMN_62288 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline