Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585140_m1
          See other GAS7 GE Assays ›
          Gene Symbol:
          Gas7
          Gene Name:
          growth arrest specific 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 54010722 - 54240798 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585140_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gas7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053484.2 NP_445936.2 8 - 9 639 88
          XM_006246827.2 XP_006246889.1 8 - 9 1308 88
          XM_006246828.2 XP_006246890.1 8 - 9 2149 88
          XM_006246829.2 XP_006246891.1 8 - 9 1492 88
          XM_006246830.2 XP_006246892.1 8 - 9 823 88
          XM_008767883.1 XP_008766105.1 8 - 9 1223 88
          GenBank mRNA AJ003148.1 - 8 - 9 775 88
          AJ131902.1 - 775 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000868.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gas7

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          regulation of cell shape
          actin filament polymerization
          cell differentiation
          neuron differentiation
          neuron projection morphogenesis
          actin filament bundle assembly
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P115781 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P177465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265447 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114117 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161968 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4553 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138737 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71095 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16296 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85980 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109858 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57832 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6314 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174893 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115781 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P177465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115781 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P177465 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P115781 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P177465 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P177465 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701283402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83545 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9794 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10527 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18833 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66331 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10480 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25927 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30201 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55735 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45681 - -
          Agilent Rat cDNA Microarray (G4105A) 701283402 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10734687 - -
          Affymetrix Rat Gene 1.0 ST Array 10734682 - -
          Affymetrix Rat Gene 1.0 ST Array 10734675 - -
          Affymetrix Rat Gene 1.0 ST Array 10734677 - -
          Affymetrix Rat Gene 1.0 ST Array 10734683 - -
          Affymetrix Rat Gene 1.0 ST Array 10734688 - -
          Affymetrix Rat Gene 1.0 ST Array 10734681 - -
          Affymetrix Rat Gene 1.0 ST Array 10734689 - -
          Affymetrix Rat Gene 1.0 ST Array 10734685 - -
          Affymetrix Rat Gene 1.0 ST Array 10734680 - -
          Affymetrix Rat Gene 1.0 ST Array 10734690 - -
          Affymetrix Rat Gene 1.0 ST Array 10734686 - -
          Affymetrix Rat Gene 1.1 ST Array 10734673 - -
          Rat Genome 230 2.0 Array 1370963_PM_at - -
          Rat Genome 230 2.0 Array 1370963_at - -
          Affymetrix HT Rat Focus Array 1370963_PM_at - -
          Affymetrix HT Rat Focus Array 1370963_at - -
          Affymetrix Rat Clariom S Assay PSR1000007394.rn.1 TC1000000868.rn.1 RTA TC
          Rat Expression Set 230 1370963_at - -
          Rat Expression Set 230 1370963_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10734673 - -
          Rat Genome U34 Set AJ131902_at - -
          Rat Genome U34 Set AJ003148cds_s_at - -
          Rat Genome U34 Set AJ131902_g_at - -
          Rat Genome U34 Set rc_AI043809_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21299235 - -
          ABI Rat Genome Survey Microarray 21586741 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356585 - -
          RatRef-12 Expression BeadChip ILMN_69714 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline