Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585167_m1
          See other PLG GE Assays ›
          Gene Symbol:
          Plg
          Gene Name:
          plasminogen
          Gene Aliases:
          Ab1-346
          Chromosome Location:
          Chr.1: 48521830 - 48563759 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585167_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053491.2 NP_445943.1 6 - 7 705 71
          GenBank mRNA AJ242649.1 - 6 - 7 708 71
          AY325159.1 - 6 - 7 657 71
          BC091135.1 - 6 - 7 705 7369357 71
          FQ210878.1 - 6 - 7 848 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004458.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Estrogen Panel Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plg

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          blood coagulation
          negative regulation of cell-substrate adhesion
          extracellular matrix disassembly
          tissue regeneration
          fibrinolysis
          myoblast differentiation
          muscle cell cellular homeostasis
          tissue remodeling
          proteolysis involved in cellular protein catabolic process
          negative regulation of fibrinolysis
          positive regulation of fibrinolysis
          trophoblast giant cell differentiation
          labyrinthine layer blood vessel development
          mononuclear cell migration
          endopeptidase activity
          serine-type endopeptidase activity
          receptor binding
          protein domain specific binding
          apolipoprotein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P599062 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136313 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59827 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116277 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P599062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017580 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P599062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017580 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017580 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P599062 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P285681 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81572 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10717979 - -
          Affymetrix Rat Gene 1.0 ST Array 10717990 - -
          Affymetrix Rat Gene 1.0 ST Array 10717981 - -
          Affymetrix Rat Gene 1.0 ST Array 10717977 - -
          Affymetrix Rat Gene 1.0 ST Array 10717988 - -
          Affymetrix Rat Gene 1.0 ST Array 10717975 - -
          Affymetrix Rat Gene 1.0 ST Array 10717968 - -
          Affymetrix Rat Gene 1.0 ST Array 10717986 - -
          Affymetrix Rat Gene 1.0 ST Array 10717982 - -
          Affymetrix Rat Gene 1.0 ST Array 10717984 - -
          Affymetrix Rat Gene 1.0 ST Array 10717973 - -
          Affymetrix Rat Gene 1.0 ST Array 10717969 - -
          Affymetrix Rat Gene 1.0 ST Array 10717970 - -
          Affymetrix Rat Gene 1.0 ST Array 10717971 - -
          Affymetrix Rat Gene 1.0 ST Array 10717991 - -
          Affymetrix Rat Gene 1.0 ST Array 10717980 - -
          Affymetrix Rat Gene 1.0 ST Array 10717987 - -
          Affymetrix Rat Gene 1.0 ST Array 10717976 - -
          Affymetrix Rat Gene 1.0 ST Array 10717972 - -
          Affymetrix Rat Gene 1.0 ST Array 10717985 - -
          Affymetrix Rat Gene 1.1 ST Array 10717967 - -
          Rat Genome 230 2.0 Array 1368360_at - -
          Rat Genome 230 2.0 Array 1368360_PM_at - -
          Affymetrix HT Rat Focus Array 1368360_PM_at - -
          Affymetrix HT Rat Focus Array 1368360_at - -
          Affymetrix Rat Clariom S Assay PSR0100035435.rn.1 TC0100004458.rn.1 RTA TC
          Rat Expression Set 230 1368360_PM_at - -
          Rat Expression Set 230 1368360_at - -
          Affymetrix Rat Gene 1.0 ST Array 10717967 - -
          Rat Genome U34 Set M62832_at - -
          Rat Genome U34 Set rc_AI072536_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21295746 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354227 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline