Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585181_m1
          See other SLC44A1 GE Assays ›
          Gene Symbol:
          Slc44a1
          Gene Name:
          solute carrier family 44 (choline transporter), member 1
          Gene Aliases:
          Cdw92, Ctl1
          Chromosome Location:
          Chr.5: 70243629 - 70424115 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585181_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc44a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033852.1 NP_001029024.1 5 - 6 514 79
          NM_053492.3 NP_445944.2 5 - 6 514 79
          XM_008763716.1 XP_008761938.1 5 - 6 547 79
          XM_008763717.1 XP_008761939.1 5 - 6 548 79
          GenBank mRNA AJ245619.1 - 5 - 6 506 79
          AJ420809.1 - 5 - 6 514 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000660.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc44a1

          Panther Classification:

          Molecular Function -

          transporter secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          choline transport
          cation transmembrane transport
          choline transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P095766 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P448710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96130 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97375 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13302 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17083 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 602 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133548 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145900 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141649 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140649 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39982 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102412 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146565 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172180 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095766 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P448710 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P448710 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095766 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P448710 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P095766 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461351 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P448710 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10969 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50583 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86356 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12375 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37551 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7122 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10740 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2654 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95200 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55128 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8788 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10692 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10869116 - -
          Affymetrix Rat Gene 1.0 ST Array 10869121 - -
          Affymetrix Rat Gene 1.0 ST Array 10869118 - -
          Affymetrix Rat Gene 1.0 ST Array 10869120 - -
          Affymetrix Rat Gene 1.0 ST Array 10869117 - -
          Affymetrix Rat Gene 1.0 ST Array 10869112 - -
          Affymetrix Rat Gene 1.0 ST Array 10869114 - -
          Affymetrix Rat Gene 1.0 ST Array 10869123 - -
          Affymetrix Rat Gene 1.0 ST Array 10869113 - -
          Affymetrix Rat Gene 1.0 ST Array 10869107 - -
          Affymetrix Rat Gene 1.0 ST Array 10869119 - -
          Affymetrix Rat Gene 1.0 ST Array 10869110 - -
          Affymetrix Rat Gene 1.0 ST Array 10869108 - -
          Affymetrix Rat Gene 1.0 ST Array 10869109 - -
          Affymetrix Rat Gene 1.0 ST Array 10869115 - -
          Affymetrix Rat Gene 1.0 ST Array 10869124 - -
          Affymetrix Rat Gene 1.0 ST Array 10869111 - -
          Affymetrix Rat Gene 1.1 ST Array 10869106 - -
          Rat Genome 230 2.0 Array 1368480_PM_at - -
          Rat Genome 230 2.0 Array 1370935_PM_at - -
          Rat Genome 230 2.0 Array 1368480_at - -
          Rat Genome 230 2.0 Array 1373054_at - -
          Rat Genome 230 2.0 Array 1370935_at - -
          Rat Genome 230 2.0 Array 1373054_PM_at - -
          Affymetrix HT Rat Focus Array 1368480_at - -
          Affymetrix HT Rat Focus Array 1370935_PM_at - -
          Affymetrix HT Rat Focus Array 1368480_PM_at - -
          Affymetrix HT Rat Focus Array 1370935_at - -
          Affymetrix HT Rat Focus Array 1373054_at - -
          Affymetrix HT Rat Focus Array 1373054_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500004481.rn.1 TC0500000660.rn.1 RTA TC
          Rat Expression Set 230 1368480_PM_at - -
          Rat Expression Set 230 1370935_at - -
          Rat Expression Set 230 1373054_at - -
          Rat Expression Set 230 1373054_PM_at - -
          Rat Expression Set 230 1368480_at - -
          Rat Expression Set 230 1370935_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10869106 - -
          Rat Genome U34 Set AA801076_at - -
          Rat Genome U34 Set rc_AI236539_at - -
          Rat Genome U34 Set rc_AI105205_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21338448 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359953 - -
          RatRef-12 Expression BeadChip ILMN_57086 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline