Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585276_m1
          See other HRH3 GE Assays ›
          Gene Symbol:
          Hrh3
          Gene Name:
          histamine receptor H3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 175474310 - 175479395 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585276_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hrh3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270564.1 NP_001257493.1 1 - 2 589 55
          NM_001270565.1 NP_001257494.1 1 - 2 589 55
          NM_001270566.1 NP_001257495.1 1 - 2 589 55
          NM_001270567.1 NP_001257496.1 1 - 2 589 55
          NM_001270568.1 NP_001257497.1 1 - 2 589 55
          NM_001270569.1 NP_001257498.1 1 - 2 589 55
          NM_001270570.1 NP_001257499.1 1 - 2 589 55
          NM_053506.2 NP_445958.1 1 - 2 589 55
          GenBank mRNA AB015646.1 - 1 - 2 589 55
          AF237919.1 - 1 - 2 252 55
          AY009370.1 - 1 - 2 252 55
          AY009371.1 - 1 - 2 252 55
          BC087707.1 - 1 - 2 587 7315203 55
          DQ112342.1 - 1 - 2 252 55
          DQ112343.1 - 1 - 2 252 55

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hrh3

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of neurotransmitter levels
          G-protein coupled receptor signaling pathway
          negative regulation of adenylate cyclase activity
          positive regulation of cytosolic calcium ion concentration
          brain development
          learning
          memory
          negative regulation of glutamate secretion
          negative regulation of gamma-aminobutyric acid secretion
          regulation of norepinephrine secretion
          negative regulation of serotonin secretion
          response to organic cyclic compound
          eating behavior
          drinking behavior
          negative regulation of blood pressure
          positive regulation of epithelial cell proliferation
          cognition
          histamine receptor activity
          drug binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P391760 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398559 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88454 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P391760 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011045 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011045 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P391760 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011045 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P391760 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P387429 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702078795 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18128 - -
          Agilent Rat cDNA Microarray (G4105A) 701745351 - -
          Agilent Rat cDNA Microarray (G4105A) 702078795 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852264 - -
          Affymetrix Rat Gene 1.0 ST Array 10852262 - -
          Affymetrix Rat Gene 1.0 ST Array 10852269 - -
          Affymetrix Rat Gene 1.0 ST Array 10852265 - -
          Affymetrix Rat Gene 1.0 ST Array 10852268 - -
          Affymetrix Rat Gene 1.0 ST Array 10852261 - -
          Affymetrix Rat Gene 1.0 ST Array 10852259 - -
          Affymetrix Rat Gene 1.0 ST Array 10852266 - -
          Affymetrix Rat Gene 1.0 ST Array 10852267 - -
          Affymetrix Rat Gene 1.0 ST Array 10852260 - -
          Affymetrix Rat Gene 1.0 ST Array 10852263 - -
          Affymetrix Rat Gene 1.1 ST Array 10852258 - -
          Rat Genome 230 2.0 Array 1368451_at - -
          Rat Genome 230 2.0 Array 1388080_PM_a_at - -
          Rat Genome 230 2.0 Array 1388080_a_at - -
          Rat Genome 230 2.0 Array 1368451_PM_at - -
          Affymetrix HT Rat Focus Array 1388080_a_at - -
          Affymetrix HT Rat Focus Array 1368451_PM_at - -
          Affymetrix HT Rat Focus Array 1388080_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368451_at - -
          Rat Expression Set 230 1388080_PM_a_at - -
          Rat Expression Set 230 1388080_a_at - -
          Rat Expression Set 230 1368451_at - -
          Rat Expression Set 230 1368451_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852258 - -
          Rat Genome U34 Set rc_AA943666_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21980719 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365804 - -
          RatRef-12 Expression BeadChip ILMN_69098 - -

          Back To Top
           

          Citations (1)

          Histaminergic responses by hypothalamic neurons that regulate lordosis and their modulation by estradiol.
          Authors
          Dupre C, Lovett-Barron M, Pfaff DW, Kow LM
          Journal
          Proc Natl Acad Sci U S A - Page No 12311 (Tue Jul 06 00:00:00 EDT 2010)
          Volume (Year)
          107 (2010)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline