Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585356_m1
          See other ELF1 GE Assays ›
          Gene Symbol:
          Elf1
          Gene Name:
          E74-like factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 61772626 - 61868442 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585356_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Elf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053520.2 NP_445972.2 1 - 2 299 98
          XM_006252347.2 XP_006252409.1 2 - 3 1558 98
          XM_006252348.2 XP_006252410.1 1 - 2 296 98
          XM_006252349.2 XP_006252411.1 3 - 4 1724 98
          XM_006252351.2 XP_006252413.1 2 - 3 802 98
          XM_006252352.2 XP_006252414.1 2 - 3 762 98
          XM_006252354.2 XP_006252416.1 2 - 3 1558 98
          XM_008770898.1 XP_008769120.1 2 - 3 3062 98
          XM_008770899.1 XP_008769121.1 2 - 3 758 98
          XM_008770900.1 XP_008769122.1 2 - 3 314 98
          XM_008770901.1 XP_008769123.1 2 - 3 3062 98
          XM_008770902.1 XP_008769124.1 2 - 3 3062 98
          XM_008770903.1 XP_008769125.1 2 - 3 3062 98
          GenBank mRNA AB030216.1 - 1 - 2 68 98
          AB030217.1 - 1 - 2 68 98
          AB046531.1 - 1 - 2 78 98
          CB782606.1 - 2 - 3 354 98
          FQ233920.1 - 2 - 3 373 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000907.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Endogenous Controls Recommended Assay Rattus norvegicus Rat Endogenous Controls 1700 Inventoried Rat_Endogenous_Controls Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Elf1

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytokine production
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of T cell receptor signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P065507 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P132050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178748 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61689 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63079 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101316 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065507 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132050 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132050 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065507 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P132050 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P065507 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12834 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95442 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54187 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65542 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27192 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781692 - -
          Affymetrix Rat Gene 1.0 ST Array 10781697 - -
          Affymetrix Rat Gene 1.0 ST Array 10781695 - -
          Affymetrix Rat Gene 1.0 ST Array 10781691 - -
          Affymetrix Rat Gene 1.0 ST Array 10781694 - -
          Affymetrix Rat Gene 1.0 ST Array 10781698 - -
          Affymetrix Rat Gene 1.0 ST Array 10781696 - -
          Affymetrix Rat Gene 1.0 ST Array 10781693 - -
          Affymetrix Rat Gene 1.1 ST Array 10781690 - -
          Rat Genome 230 2.0 Array 1374137_PM_at - -
          Rat Genome 230 2.0 Array 1374137_at - -
          Rat Genome 230 2.0 Array 1370542_a_at - -
          Rat Genome 230 2.0 Array 1370542_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370542_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370542_a_at - -
          Affymetrix Rat Clariom S Assay PSR1500006265.rn.1 TC1500000907.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500006263.rn.1 TC1500000907.rn.1 RTA TC
          Rat Expression Set 230 1370542_PM_a_at - -
          Rat Expression Set 230 1370542_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781690 - -
          Rat Genome U34 Set rc_AA944441_at - -
          Rat Genome U34 Set rc_AA850537_at - -
          Rat Genome U34 Set rc_AA850537_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22375118 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372256 - -
          RatRef-12 Expression BeadChip ILMN_56835 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline