Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585478_m1
          See other VAMP7 GE Assays ›
          Gene Symbol:
          Vamp7
          Gene Name:
          vesicle-associated membrane protein 7
          Gene Aliases:
          Sybl1
          Chromosome Location:
          Chr.12: 18996566 - 19032330 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585478_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vamp7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053531.1 NP_445983.1 5 - 6 502 93
          XM_006249016.2 XP_006249078.1 6 - 7 655 93
          GenBank mRNA AF281632.1 - 5 - 6 502 93
          FQ228923.1 - 6 - 7 615 93
          FQ230649.1 - 5 - 6 426 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000317.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vamp7

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          ER to Golgi vesicle-mediated transport
          vesicle fusion
          phagocytosis, engulfment
          endosome to lysosome transport
          vesicle-mediated transport
          calcium ion regulated exocytosis
          triglyceride transport
          SNARE complex assembly
          Golgi to plasma membrane protein transport
          eosinophil degranulation
          neutrophil degranulation
          natural killer cell degranulation
          vesicle transport along microtubule
          vesicle fusion with Golgi apparatus
          positive regulation of dendrite morphogenesis
          autophagosome maturation
          regulation of protein targeting to vacuolar membrane
          positive regulation of histamine secretion by mast cell
          SNAP receptor activity
          protein binding
          syntaxin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96955 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12839 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12839 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12839 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12839 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5043 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102957 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10760803 - -
          Affymetrix Rat Gene 1.0 ST Array 10760800 - -
          Affymetrix Rat Gene 1.0 ST Array 10760802 - -
          Affymetrix Rat Gene 1.0 ST Array 10760798 - -
          Affymetrix Rat Gene 1.0 ST Array 10760804 - -
          Affymetrix Rat Gene 1.0 ST Array 10760799 - -
          Affymetrix Rat Gene 1.0 ST Array 10760801 - -
          Affymetrix Rat Gene 1.1 ST Array 10760797 - -
          Rat Genome 230 2.0 Array 1370098_at - -
          Rat Genome 230 2.0 Array 1392903_at - -
          Rat Genome 230 2.0 Array 1379996_at - -
          Rat Genome 230 2.0 Array 1379996_PM_at - -
          Rat Genome 230 2.0 Array 1370098_PM_at - -
          Rat Genome 230 2.0 Array 1392903_PM_at - -
          Affymetrix HT Rat Focus Array 1370098_PM_at - -
          Affymetrix HT Rat Focus Array 1370098_at - -
          Affymetrix Rat Clariom S Assay PSR1200002534.rn.1 TC1200000317.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200002536.rn.1 TC1200000317.rn.1 RTA TC
          Rat Expression Set 230 1370098_PM_at - -
          Rat Expression Set 230 1370098_at - -
          Affymetrix Rat Gene 1.0 ST Array 10760797 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21955106 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352570 - -
          RatRef-12 Expression BeadChip ILMN_50581 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline