Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585521_m1
          See other LAPTM5 GE Assays ›
          Gene Symbol:
          Laptm5
          Gene Name:
          lysosomal protein transmembrane 5
          Gene Aliases:
          gcd-10
          Chromosome Location:
          Chr.5: 149047681 - 149069719 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585521_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Laptm5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053538.2 NP_445990.2 3 - 4 262 63
          GenBank mRNA AB046592.1 - 3 - 4 274 63
          BC060517.1 - 3 - 4 262 5597893 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001570.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Laptm5

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s)

          transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97952 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45369 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11236 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11236 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11236 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11236 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97200 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20406 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8816 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872497 - -
          Affymetrix Rat Gene 1.0 ST Array 10872493 - -
          Affymetrix Rat Gene 1.0 ST Array 10872498 - -
          Affymetrix Rat Gene 1.0 ST Array 10872494 - -
          Affymetrix Rat Gene 1.0 ST Array 10872496 - -
          Affymetrix Rat Gene 1.0 ST Array 10872490 - -
          Affymetrix Rat Gene 1.0 ST Array 10872492 - -
          Affymetrix Rat Gene 1.0 ST Array 10872500 - -
          Affymetrix Rat Gene 1.0 ST Array 10872495 - -
          Affymetrix Rat Gene 1.0 ST Array 10872499 - -
          Affymetrix Rat Gene 1.1 ST Array 10872489 - -
          Rat Genome 230 2.0 Array 1383658_PM_at - -
          Rat Genome 230 2.0 Array 1368006_PM_at - -
          Rat Genome 230 2.0 Array 1383658_at - -
          Rat Genome 230 2.0 Array 1368006_at - -
          Affymetrix HT Rat Focus Array 1368006_PM_at - -
          Affymetrix HT Rat Focus Array 1383658_PM_at - -
          Affymetrix HT Rat Focus Array 1383658_at - -
          Affymetrix HT Rat Focus Array 1368006_at - -
          Affymetrix Rat Clariom S Assay PSR0500011328.rn.1 TC0500001570.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011333.rn.1 TC0500001570.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011329.rn.1 TC0500001570.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011330.rn.1 TC0500001570.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011327.rn.1 TC0500001570.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011331.rn.1 TC0500001570.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011334.rn.1 TC0500001570.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011337.rn.1 TC0500001570.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011335.rn.1 TC0500001570.rn.1 RTA TC
          Rat Expression Set 230 1368006_at - -
          Rat Expression Set 230 1368006_PM_at - -
          Rat Expression Set 230 1383658_PM_at - -
          Rat Expression Set 230 1383658_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872489 - -
          Rat Genome U34 Set rc_AA925353_at - -
          Rat Genome U34 Set rc_AI234133_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21455435 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357030 - -
          RatRef-12 Expression BeadChip ILMN_49562 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline