Search
Search
| Gene Symbol: Idi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053539.2 | NP_445991.2 | 1 - 2 | 372 | 88 | |
| GenBank mRNA | AF003835.1 | - | 1 - 2 | 88 | ||
| BC089786.1 | - | 1 - 2 | 372 | 7322086 | 88 | |
Set Membership: |
|