Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585549_m1
          See other SFRP4 GE Assays ›
          Gene Symbol:
          Sfrp4
          Gene Name:
          secreted frizzled-related protein 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 47383983 - 47394065 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585549_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sfrp4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053544.1 NP_445996.1 5 - 6 948 101
          GenBank mRNA AF012891.1 - 5 - 6 948 101
          AF140346.1 - 5 - 6 1099 101
          FQ220237.1 - 2 - 3 257 101
          FQ229078.1 - 2 - 3 270 101
          FQ229224.1 - 4 - 5 379 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001868.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Rat Wnt Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sfrp4

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor internalization
          multicellular organism development
          female pregnancy
          negative regulation of cell proliferation
          positive regulation of gene expression
          Wnt signaling pathway
          cell differentiation
          response to estradiol
          non-canonical Wnt signaling pathway
          positive regulation of apoptotic process
          negative regulation of sequence-specific DNA binding transcription factor activity
          response to peptide hormone
          positive regulation of epidermal cell differentiation
          negative regulation of JNK cascade
          decidualization
          response to glucocorticoid
          phosphate ion homeostasis
          mammary gland involution
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          positive regulation of keratinocyte apoptotic process
          negative regulation of sodium-dependent phosphate transport
          G-protein coupled receptor activity
          Wnt-protein binding
          Wnt-activated receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P290187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40281 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87236 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67424 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P290187 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P290187 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P290187 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P292745 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16129 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P290187 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31983 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99352 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57712 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798614 - -
          Affymetrix Rat Gene 1.0 ST Array 10798618 - -
          Affymetrix Rat Gene 1.0 ST Array 10798620 - -
          Affymetrix Rat Gene 1.0 ST Array 10798613 - -
          Affymetrix Rat Gene 1.0 ST Array 10798611 - -
          Affymetrix Rat Gene 1.0 ST Array 10798617 - -
          Affymetrix Rat Gene 1.0 ST Array 10798615 - -
          Affymetrix Rat Gene 1.0 ST Array 10798619 - -
          Affymetrix Rat Gene 1.0 ST Array 10798616 - -
          Affymetrix Rat Gene 1.0 ST Array 10798612 - -
          Affymetrix Rat Gene 1.1 ST Array 10798610 - -
          Rat Genome 230 2.0 Array 1368394_PM_at - -
          Rat Genome 230 2.0 Array 1388021_PM_at - -
          Rat Genome 230 2.0 Array 1368394_at - -
          Rat Genome 230 2.0 Array 1388021_at - -
          Affymetrix HT Rat Focus Array 1388021_at - -
          Affymetrix HT Rat Focus Array 1388021_PM_at - -
          Affymetrix HT Rat Focus Array 1368394_PM_at - -
          Affymetrix HT Rat Focus Array 1368394_at - -
          Affymetrix Rat Clariom S Assay PSR1700012709.rn.1 TC1700001868.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700012710.rn.1 TC1700001868.rn.1 RTA TC
          Rat Expression Set 230 1368394_PM_at - -
          Rat Expression Set 230 1388021_PM_at - -
          Rat Expression Set 230 1388021_at - -
          Rat Expression Set 230 1368394_at - -
          Affymetrix Rat Gene 1.0 ST Array 10798610 - -
          Rat Genome U34 Set AF012891_at - -
          Rat Genome U34 Set AF012891_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21603783 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352716 - -
          RatRef-12 Expression BeadChip ILMN_55615 - -

          Back To Top
           

          Citations (1)

          Catch-up growth after hypothyroidism is caused by delayed growth plate senescence.
          Authors
          Marino R, Hegde A, Barnes KM, Schrier L, Emons JA, Nilsson O, Baron J
          Journal
          Endocrinology - Page No 1820 (Tue Apr 01 00:00:00 EDT 2008)
          Volume (Year)
          149 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline