Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585577_m1
          See other PDK4 GE Assays ›
          Gene Symbol:
          Pdk4
          Gene Name:
          pyruvate dehydrogenase kinase, isozyme 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 30546772 - 30556774 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585577_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdk4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053551.1 NP_446003.1 6 - 7 800 76
          GenBank mRNA AF034577.1 - 6 - 7 800 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002408.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdk4

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          acetyl-CoA biosynthetic process from pyruvate
          protein phosphorylation
          regulation of pH
          insulin receptor signaling pathway
          cellular response to starvation
          regulation of acetyl-CoA biosynthetic process from pyruvate
          regulation of cellular ketone metabolic process
          regulation of glucose metabolic process
          regulation of fatty acid biosynthetic process
          glucose homeostasis
          response to starvation
          regulation of bone resorption
          regulation of fatty acid oxidation
          cellular response to fatty acid
          reactive oxygen species metabolic process
          negative regulation of anoikis
          protein kinase activity
          pyruvate dehydrogenase (acetyl-transferring) kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P231269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171697 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P231269 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P231269 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P231269 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P231269 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700187344 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56169 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16093 - -
          Agilent Rat cDNA Microarray (G4105A) 700187344 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10860906 - -
          Affymetrix Rat Gene 1.0 ST Array 10860907 - -
          Affymetrix Rat Gene 1.0 ST Array 10860911 - -
          Affymetrix Rat Gene 1.0 ST Array 10860910 - -
          Affymetrix Rat Gene 1.0 ST Array 10860905 - -
          Affymetrix Rat Gene 1.0 ST Array 10860903 - -
          Affymetrix Rat Gene 1.0 ST Array 10860901 - -
          Affymetrix Rat Gene 1.0 ST Array 10860904 - -
          Affymetrix Rat Gene 1.0 ST Array 10860902 - -
          Affymetrix Rat Gene 1.0 ST Array 10860909 - -
          Affymetrix Rat Gene 1.0 ST Array 10860908 - -
          Affymetrix Rat Gene 1.1 ST Array 10860900 - -
          Rat Genome 230 2.0 Array 1369150_at - -
          Rat Genome 230 2.0 Array 1369150_PM_at - -
          Affymetrix HT Rat Focus Array 1369150_PM_at - -
          Affymetrix HT Rat Focus Array 1369150_at - -
          Affymetrix Rat Clariom S Assay PSR0400017215.rn.1 TC0400002408.rn.1 RTA TC
          Rat Expression Set 230 1369150_PM_at - -
          Rat Expression Set 230 1369150_at - -
          Affymetrix Rat Gene 1.0 ST Array 10860900 - -
          Rat Genome U34 Set AF034577_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21936440 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364094 - -
          RatRef-12 Expression BeadChip ILMN_60681 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline