Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585606_m1
          See other PRMT3 GE Assays ›
          Gene Symbol:
          Prmt3
          Gene Name:
          protein arginine methyltransferase 3
          Gene Aliases:
          Hrmt1l3
          Chromosome Location:
          Chr.1: 105113595 - 105200253 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585606_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prmt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053557.1 NP_446009.1 13 - 14 1418 110
          GenBank mRNA AF059530.1 - 13 - 14 1418 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001556.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prmt3

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          protein methylation
          peptidyl-arginine methylation
          peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
          negative regulation of protein ubiquitination
          histone arginine methylation
          dendritic spine morphogenesis
          methyltransferase activity
          histone-arginine N-methyltransferase activity
          S-adenosylmethionine-dependent methyltransferase activity
          protein-arginine N-methyltransferase activity
          protein-arginine omega-N asymmetric methyltransferase activity
          metal ion binding
          modified amino acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P536125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707919 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160879 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103930 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18578 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49351 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92608 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112115 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21648 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161514 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P536125 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P536125 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P536125 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P536125 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700415191 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2440 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57025 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98753 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72978 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91638 - -
          Agilent Rat cDNA Microarray (G4105A) 700415191 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10707241 - -
          Affymetrix Rat Gene 1.0 ST Array 10707245 - -
          Affymetrix Rat Gene 1.0 ST Array 10707249 - -
          Affymetrix Rat Gene 1.0 ST Array 10707247 - -
          Affymetrix Rat Gene 1.0 ST Array 10707246 - -
          Affymetrix Rat Gene 1.0 ST Array 10707243 - -
          Affymetrix Rat Gene 1.0 ST Array 10707238 - -
          Affymetrix Rat Gene 1.0 ST Array 10707251 - -
          Affymetrix Rat Gene 1.0 ST Array 10707244 - -
          Affymetrix Rat Gene 1.0 ST Array 10707240 - -
          Affymetrix Rat Gene 1.0 ST Array 10707242 - -
          Affymetrix Rat Gene 1.0 ST Array 10707253 - -
          Affymetrix Rat Gene 1.0 ST Array 10707239 - -
          Affymetrix Rat Gene 1.0 ST Array 10707252 - -
          Affymetrix Rat Gene 1.0 ST Array 10707250 - -
          Affymetrix Rat Gene 1.0 ST Array 10707248 - -
          Affymetrix Rat Gene 1.1 ST Array 10707237 - -
          Rat Genome 230 2.0 Array 1370536_at - -
          Rat Genome 230 2.0 Array 1370536_PM_at - -
          Affymetrix HT Rat Focus Array 1370536_at - -
          Affymetrix HT Rat Focus Array 1370536_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100012595.rn.1 TC0100001556.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012589.rn.1 TC0100001556.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012590.rn.1 TC0100001556.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012591.rn.1 TC0100001556.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012594.rn.1 TC0100001556.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012592.rn.1 TC0100001556.rn.1 RTA TC
          Rat Expression Set 230 1370536_at - -
          Rat Expression Set 230 1370536_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10707237 - -
          Rat Genome U34 Set AF059530_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21000121 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373328 - -
          RatRef-12 Expression BeadChip ILMN_50056 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline