Search
Search
| Gene Symbol: Nap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053561.2 | NP_446013.2 | 7 - 8 | 725 | 123 | |
| XM_006241342.1 | XP_006241404.1 | 7 - 8 | 739 | 123 | ||
| XM_006241343.2 | XP_006241405.1 | 7 - 8 | 739 | 123 | ||
| XM_006241344.2 | XP_006241406.1 | 5 - 6 | 476 | 123 | ||
| GenBank mRNA | AF062594.1 | - | 7 - 8 | 707 | 120 | |
| CF110129.1 | - | 2 - 3 | 160 | 123 | ||
Set Membership: |
|