Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585690_m1
          See other FTCD GE Assays ›
          Gene Symbol:
          Ftcd
          Gene Name:
          formimidoyltransferase cyclodeaminase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 12806957 - 12820466 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585690_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ftcd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053567.1 NP_446019.1 7 - 8 927 87
          GenBank mRNA AF079233.3 - 7 - 8 927 87
          BC091134.1 - 7 - 8 939 7369085 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001326.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ftcd

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          histidine catabolic process to glutamate and formamide
          histidine catabolic process to glutamate and formate
          tetrahydrofolate interconversion
          folic acid binding
          intermediate filament binding
          glutamate formimidoyltransferase activity
          formimidoyltetrahydrofolate cyclodeaminase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P992662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120654 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P992662 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P992662 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P992662 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P992662 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53696 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10832359 - -
          Affymetrix Rat Gene 1.0 ST Array 10832362 - -
          Affymetrix Rat Gene 1.0 ST Array 10832361 - -
          Affymetrix Rat Gene 1.0 ST Array 10832367 - -
          Affymetrix Rat Gene 1.0 ST Array 10832360 - -
          Affymetrix Rat Gene 1.0 ST Array 10832369 - -
          Affymetrix Rat Gene 1.0 ST Array 10832370 - -
          Affymetrix Rat Gene 1.0 ST Array 10832371 - -
          Affymetrix Rat Gene 1.0 ST Array 10832363 - -
          Affymetrix Rat Gene 1.0 ST Array 10832366 - -
          Affymetrix Rat Gene 1.0 ST Array 10832372 - -
          Affymetrix Rat Gene 1.0 ST Array 10832368 - -
          Affymetrix Rat Gene 1.0 ST Array 10832364 - -
          Affymetrix Rat Gene 1.0 ST Array 10832365 - -
          Affymetrix Rat Gene 1.1 ST Array 10796785 - -
          Affymetrix Rat Gene 1.1 ST Array 10832358 - -
          Affymetrix Rat Gene 1.1 ST Array 10874909 - -
          Rat Genome 230 2.0 Array 1387877_at - -
          Rat Genome 230 2.0 Array 1387877_PM_at - -
          Affymetrix HT Rat Focus Array 1387877_PM_at - -
          Affymetrix HT Rat Focus Array 1387877_at - -
          Affymetrix Rat Clariom S Assay PSR2000010913.rn.1 TC2000001326.rn.1 RTA TC
          Rat Expression Set 230 1387877_at - -
          Rat Expression Set 230 1387877_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10832358 - -
          Rat Genome U34 Set rc_AI072384_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21655467 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349772 - -
          RatRef-12 Expression BeadChip ILMN_69833 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline