Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585883_m1
          See other RAB14 GE Assays ›
          Gene Symbol:
          Rab14
          Gene Name:
          RAB14, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 14373270 - 14395357 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585883_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053589.1 NP_446041.1 4 - 5 354 77
          XM_006234049.2 XP_006234111.1 5 - 6 787 77
          XM_006234050.2 XP_006234112.1 5 - 6 439 77
          GenBank mRNA BC158585.1 - 4 - 5 943 7446356 77
          M83680.1 - 4 - 5 354 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002929.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab14

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          Golgi to endosome transport
          small GTPase mediated signal transduction
          body fluid secretion
          metabolic process
          fibroblast growth factor receptor signaling pathway
          protein transport
          vesicle-mediated transport
          endocytic recycling
          Rab protein signal transduction
          regulation of protein localization
          apical protein localization
          intracellular transport
          phagolysosome assembly involved in apoptotic cell clearance
          glycoprotein binding
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          myosin V binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P205883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23561 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177955 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P205883 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P205883 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P205883 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P205887 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P205883 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700181949 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700181741 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35300 - -
          Agilent Rat cDNA Microarray (G4105A) 700181741 - -
          Agilent Rat cDNA Microarray (G4105A) 700181949 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10844664 - -
          Affymetrix Rat Gene 1.0 ST Array 10844660 - -
          Affymetrix Rat Gene 1.0 ST Array 10844662 - -
          Affymetrix Rat Gene 1.0 ST Array 10844661 - -
          Affymetrix Rat Gene 1.0 ST Array 10844665 - -
          Affymetrix Rat Gene 1.0 ST Array 10844659 - -
          Affymetrix Rat Gene 1.0 ST Array 10844663 - -
          Affymetrix Rat Gene 1.1 ST Array 10844658 - -
          Rat Genome 230 2.0 Array 1375687_at - -
          Rat Genome 230 2.0 Array 1375687_PM_at - -
          Rat Genome 230 2.0 Array 1385314_PM_at - -
          Rat Genome 230 2.0 Array 1397463_PM_at - -
          Rat Genome 230 2.0 Array 1391599_at - -
          Rat Genome 230 2.0 Array 1376796_PM_at - -
          Rat Genome 230 2.0 Array 1385314_at - -
          Rat Genome 230 2.0 Array 1376796_at - -
          Rat Genome 230 2.0 Array 1367884_at - -
          Rat Genome 230 2.0 Array 1391599_PM_at - -
          Rat Genome 230 2.0 Array 1367884_PM_at - -
          Rat Genome 230 2.0 Array 1397463_at - -
          Affymetrix HT Rat Focus Array 1367884_PM_at - -
          Affymetrix HT Rat Focus Array 1367884_at - -
          Affymetrix Rat Clariom S Assay PSR0300022857.rn.1 TC0300002929.rn.1 RTA TC
          Rat Expression Set 230 1367884_PM_at - -
          Rat Expression Set 230 1376796_PM_at - -
          Rat Expression Set 230 1367884_at - -
          Rat Expression Set 230 1376796_at - -
          Rat Expression Set 230 1397463_PM_at - -
          Rat Expression Set 230 1397463_at - -
          Affymetrix Rat Gene 1.0 ST Array 10844658 - -
          Rat Genome U34 Set M83680_at - -
          Rat Genome U34 Set rc_AA875010_at - -
          Rat Genome U34 Set rc_AA923974_at - -
          Rat Genome U34 Set rc_AI172532_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20733304 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348958 - -
          RatRef-12 Expression BeadChip ILMN_50327 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline