Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585886_m1
          See other PSMB1 GE Assays ›
          Gene Symbol:
          Psmb1
          Gene Name:
          proteasome (prosome, macropain) subunit, beta type 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 57470878 - 57491359 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585886_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053590.1 NP_446042.1 1 - 2 122 87
          XM_006227981.1 XP_006228043.1 3 - 4 377 87
          GenBank mRNA BC058455.1 - 1 - 2 134 6922317 87
          FQ211516.1 - 1 - 2 137 87
          FQ214442.1 - 1 - 2 137 87
          FQ214726.1 - 3 - 4 377 87
          FQ225328.1 - 1 - 2 137 87
          FQ229756.1 - 1 - 2 137 87
          FQ230094.1 - 1 - 2 137 87
          X52783.1 - 1 - 2 122 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004575.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmb1

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis involved in cellular protein catabolic process
          threonine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P130559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401392 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52790 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113339 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P130559 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P130559 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P130559 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P130559 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87562 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718299 - -
          Affymetrix Rat Gene 1.0 ST Array 10718300 - -
          Affymetrix Rat Gene 1.0 ST Array 10718302 - -
          Affymetrix Rat Gene 1.0 ST Array 10718298 - -
          Affymetrix Rat Gene 1.0 ST Array 10718301 - -
          Affymetrix Rat Gene 1.0 ST Array 10718297 - -
          Affymetrix Rat Gene 1.1 ST Array 10718296 - -
          Rat Genome 230 2.0 Array 1398812_PM_at - -
          Rat Genome 230 2.0 Array 1398812_at - -
          Affymetrix HT Rat Focus Array 1398812_at - -
          Affymetrix HT Rat Focus Array 1398812_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100036236.rn.1 TC0100004575.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036239.rn.1 TC0100004575.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036240.rn.1 TC0100004575.rn.1 RTA TC
          Rat Expression Set 230 1398812_at - -
          Rat Expression Set 230 1398812_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10718296 - -
          Rat Genome U34 Set rc_AA849722_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21963055 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357406 - -
          RatRef-12 Expression BeadChip ILMN_66471 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline