Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585893_m1
          See other DPEP1 GE Assays ›
          Gene Symbol:
          Dpep1
          Gene Name:
          dipeptidase 1 (renal)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 55973458 - 55998830 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585893_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dpep1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053591.2 NP_446043.1 2 - 3 253 80
          XM_006255783.2 XP_006255845.1 2 - 3 248 80
          GenBank mRNA BC072476.1 - 2 - 3 253 7100683 80
          L07315.1 - 2 - 3 231 80
          L07316.1 - 2 - 3 231 80
          M94056.1 - 2 - 3 231 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000799.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dpep1

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          antibiotic metabolic process
          negative regulation of cell migration
          cellular response to drug
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          homocysteine metabolic process
          cellular response to calcium ion
          cellular response to nitric oxide
          dipeptidyl-peptidase activity
          zinc ion binding
          dipeptidase activity
          GPI anchor binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          metallodipeptidase activity
          modified amino acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P009212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96947 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134953 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009212 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009212 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P009212 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P819194 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702142102 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72693 - -
          Agilent Rat cDNA Microarray (G4105A) 702142102 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10808622 - -
          Affymetrix Rat Gene 1.0 ST Array 10808629 - -
          Affymetrix Rat Gene 1.0 ST Array 10808632 - -
          Affymetrix Rat Gene 1.0 ST Array 10808624 - -
          Affymetrix Rat Gene 1.0 ST Array 10808628 - -
          Affymetrix Rat Gene 1.0 ST Array 10808631 - -
          Affymetrix Rat Gene 1.0 ST Array 10808625 - -
          Affymetrix Rat Gene 1.0 ST Array 10808621 - -
          Affymetrix Rat Gene 1.0 ST Array 10808623 - -
          Affymetrix Rat Gene 1.0 ST Array 10808626 - -
          Affymetrix Rat Gene 1.0 ST Array 10808630 - -
          Affymetrix Rat Gene 1.0 ST Array 10808627 - -
          Affymetrix Rat Gene 1.1 ST Array 10808620 - -
          Rat Genome 230 2.0 Array 1368282_PM_at - -
          Rat Genome 230 2.0 Array 1368282_at - -
          Rat Genome 230 2.0 Array 1368281_PM_at - -
          Rat Genome 230 2.0 Array 1368281_at - -
          Affymetrix HT Rat Focus Array 1368282_at - -
          Affymetrix HT Rat Focus Array 1368282_PM_at - -
          Affymetrix HT Rat Focus Array 1368281_PM_at - -
          Affymetrix HT Rat Focus Array 1368281_at - -
          Affymetrix Rat Clariom S Assay PSR1900006467.rn.1 TC1900000799.rn.1 RTA TC
          Rat Expression Set 230 1368281_at - -
          Rat Expression Set 230 1368281_PM_at - -
          Rat Expression Set 230 1368282_PM_at - -
          Rat Expression Set 230 1368282_at - -
          Affymetrix Rat Gene 1.0 ST Array 10808620 - -
          Rat Genome U34 Set L07315_at - -
          Rat Genome U34 Set rc_AI170411_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21336957 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373724 - -
          RatRef-12 Expression BeadChip ILMN_50961 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline