Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585902_m1
          See other DUT GE Assays ›
          Gene Symbol:
          Dut
          Gene Name:
          deoxyuridine triphosphatase
          Gene Aliases:
          Dutp, PIP4
          Chromosome Location:
          Chr.3: 117514401 - 117525450 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585902_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dut
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001040271.1 NP_001035361.1 1 - 2 163 148
          NM_053592.2 NP_446044.1 2 - 3 283 148
          XM_008762188.1 XP_008760410.1 2 - 3 321 148
          GenBank mRNA AW530495.1 - 1 - 2 171 148
          CB325379.1 - 1 - 2 177 148
          FQ220451.1 - 318 148
          U64030.2 - 2 - 3 283 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001625.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dut

          Panther Classification:

          Molecular Function -

          hydrolase phosphatase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          dUMP biosynthetic process
          response to organic cyclic compound
          regulation of protein heterodimerization activity
          dUTP catabolic process
          protein homotrimerization
          negative regulation of receptor activity
          magnesium ion binding
          dUTP diphosphatase activity
          receptor inhibitor activity
          pyrimidine deoxyribonucleotide binding
          peroxisome proliferator activated receptor binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P290519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932462 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29618 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P290519 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P290519 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P290519 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P290519 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12863 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700815920 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29797 - -
          Agilent Rat cDNA Microarray (G4105A) 700815920 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839407 - -
          Affymetrix Rat Gene 1.0 ST Array 10839412 - -
          Affymetrix Rat Gene 1.0 ST Array 10839408 - -
          Affymetrix Rat Gene 1.0 ST Array 10717994 - -
          Affymetrix Rat Gene 1.0 ST Array 10839410 - -
          Affymetrix Rat Gene 1.0 ST Array 10839406 - -
          Affymetrix Rat Gene 1.0 ST Array 10839409 - -
          Affymetrix Rat Gene 1.0 ST Array 10839411 - -
          Affymetrix Rat Gene 1.0 ST Array 10717993 - -
          Affymetrix Rat Gene 1.1 ST Array 10717992 - -
          Affymetrix Rat Gene 1.1 ST Array 10839405 - -
          Rat Genome 230 2.0 Array 1377424_PM_at - -
          Rat Genome 230 2.0 Array 1395361_PM_at - -
          Rat Genome 230 2.0 Array 1395361_at - -
          Rat Genome 230 2.0 Array 1377424_at - -
          Rat Genome 230 2.0 Array 1387865_at - -
          Rat Genome 230 2.0 Array 1387865_PM_at - -
          Affymetrix HT Rat Focus Array 1377424_PM_at - -
          Affymetrix HT Rat Focus Array 1387865_at - -
          Affymetrix HT Rat Focus Array 1377424_at - -
          Affymetrix HT Rat Focus Array 1395361_PM_at - -
          Affymetrix HT Rat Focus Array 1395361_at - -
          Affymetrix HT Rat Focus Array 1387865_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300011416.rn.1 TC0300001625.rn.1 RTA TC
          Rat Expression Set 230 1387865_at - -
          Rat Expression Set 230 1387865_PM_at - -
          Rat Expression Set 230 1377424_at - -
          Rat Expression Set 230 1377424_PM_at - -
          Rat Expression Set 230 1395361_at - -
          Rat Expression Set 230 1395361_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10839405 - -
          Rat Genome U34 Set rc_H31695_at - -
          Rat Genome U34 Set U64030_at - -
          Rat Genome U34 Set U64030_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21784069 - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_67201 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline