Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586037_m1
          See other BAG6 GE Assays ›
          Gene Symbol:
          Bag6
          Gene Name:
          BCL2-associated athanogene 6
          Gene Aliases:
          Bat3
          Chromosome Location:
          Chr.20: 5125324 - 5138084 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586037_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bag6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033968.1 NP_001029140.1 6 - 7 652 83
          NM_053609.2 NP_446061.2 6 - 7 810 83
          XM_006256082.1 XP_006256144.1 6 - 7 714 83
          XM_006256083.1 XP_006256145.1 6 - 7 714 83
          XM_006256084.1 XP_006256146.1 5 - 6 660 83
          XM_006256085.1 XP_006256147.1 6 - 7 714 83
          XM_006256086.2 XP_006256148.1 6 - 7 850 83
          XM_006256087.2 XP_006256149.1 6 - 7 927 83
          XM_006256088.1 XP_006256150.1 6 - 7 714 83
          XM_006256089.1 XP_006256151.1 6 - 7 714 83
          XM_006256090.1 XP_006256152.1 5 - 6 660 83
          XM_006256091.1 XP_006256153.1 5 - 6 660 83
          XM_006256092.1 XP_006256154.1 6 - 7 714 83
          XM_006256093.2 XP_006256155.1 5 - 6 798 83
          XM_006256094.1 XP_006256156.1 6 - 7 714 83
          GenBank mRNA AB018791.1 - 6 - 7 83
          BC100141.1 - 6 - 7 652 7315623 83
          FQ223705.1 - 5 - 6 773 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000197.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bag6

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          immune system process
          ubiquitin-dependent protein catabolic process
          transport
          synaptonemal complex assembly
          spermatogenesis
          brain development
          chromatin modification
          internal peptidyl-lysine acetylation
          cell differentiation
          lung development
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of proteolysis
          protein stabilization
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          tail-anchored membrane protein insertion into ER membrane
          Hsp70 protein binding
          polyubiquitin binding
          ubiquitin protein ligase binding
          ribosome binding
          proteasome binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P506345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P506345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P506345 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P506345 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P506345 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700330135 - -
          Agilent Rat cDNA Microarray (G4105A) 700330135 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828080 - -
          Affymetrix Rat Gene 1.0 ST Array 10831193 - -
          Affymetrix Rat Gene 1.0 ST Array 10831191 - -
          Affymetrix Rat Gene 1.0 ST Array 10831180 - -
          Affymetrix Rat Gene 1.0 ST Array 10831181 - -
          Affymetrix Rat Gene 1.0 ST Array 10831172 - -
          Affymetrix Rat Gene 1.0 ST Array 10831169 - -
          Affymetrix Rat Gene 1.0 ST Array 10831182 - -
          Affymetrix Rat Gene 1.0 ST Array 10831178 - -
          Affymetrix Rat Gene 1.0 ST Array 10831175 - -
          Affymetrix Rat Gene 1.0 ST Array 10831179 - -
          Affymetrix Rat Gene 1.0 ST Array 10831183 - -
          Affymetrix Rat Gene 1.0 ST Array 10831189 - -
          Affymetrix Rat Gene 1.0 ST Array 10831185 - -
          Affymetrix Rat Gene 1.0 ST Array 10831194 - -
          Affymetrix Rat Gene 1.0 ST Array 10831176 - -
          Affymetrix Rat Gene 1.0 ST Array 10831174 - -
          Affymetrix Rat Gene 1.0 ST Array 10831192 - -
          Affymetrix Rat Gene 1.0 ST Array 10831188 - -
          Affymetrix Rat Gene 1.0 ST Array 10831186 - -
          Affymetrix Rat Gene 1.0 ST Array 10831190 - -
          Affymetrix Rat Gene 1.0 ST Array 10831170 - -
          Affymetrix Rat Gene 1.0 ST Array 10831187 - -
          Affymetrix Rat Gene 1.0 ST Array 10831195 - -
          Affymetrix Rat Gene 1.0 ST Array 10831173 - -
          Affymetrix Rat Gene 1.0 ST Array 10831177 - -
          Affymetrix Rat Gene 1.0 ST Array 10831171 - -
          Affymetrix Rat Gene 1.0 ST Array 10831196 - -
          Affymetrix Rat Gene 1.1 ST Array 10831168 - -
          Rat Genome 230 2.0 Array 1387175_PM_a_at - -
          Rat Genome 230 2.0 Array 1375021_PM_at - -
          Rat Genome 230 2.0 Array 1387175_a_at - -
          Rat Genome 230 2.0 Array 1375323_a_at - -
          Rat Genome 230 2.0 Array 1375021_at - -
          Rat Genome 230 2.0 Array 1375323_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387175_PM_a_at - -
          Affymetrix HT Rat Focus Array 1375323_a_at - -
          Affymetrix HT Rat Focus Array 1375323_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387175_a_at - -
          Affymetrix Rat Clariom S Assay PSR2000001901.rn.1 TC2000000197.rn.1 RTA TC
          Rat Expression Set 230 1375323_a_at - -
          Rat Expression Set 230 1387175_a_at - -
          Rat Expression Set 230 1375323_PM_a_at - -
          Rat Expression Set 230 1387175_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10831168 - -
          Rat Genome U34 Set rc_AI010293_at - -
          Rat Genome U34 Set rc_AI008247_at - -
          Rat Genome U34 Set rc_AA944698_at - -
          Rat Genome U34 Set rc_AI010293_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22280825 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358540 - -
          RatRef-12 Expression BeadChip ILMN_59676 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0