Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586040_m1
          See other PRDX5 GE Assays ›
          Gene Symbol:
          Prdx5
          Gene Name:
          peroxiredoxin 5
          Gene Aliases:
          Aoeb166
          Chromosome Location:
          Chr.1: 222164462 - 222167447 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586040_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdx5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053610.1 NP_446062.1 1 - 2 213 69
          GenBank mRNA AF110732.1 - 1 - 2 213 69
          BC078771.1 - 1 - 2 189 7193191 69
          FQ210686.1 - 1 - 2 144 69
          FQ223238.1 - 1 - 2 141 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100008151.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdx5

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to oxidative stress
          negative regulation of transcription from RNA polymerase III promoter
          positive regulation of collagen biosynthetic process
          cellular response to reactive oxygen species
          hydrogen peroxide catabolic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of oxidoreductase activity
          oxidation-reduction process
          regulation of apoptosis involved in tissue homeostasis
          NADPH oxidation
          reactive nitrogen species metabolic process
          RNA polymerase III regulatory region DNA binding
          peroxidase activity
          receptor binding
          antioxidant activity
          oxidoreductase activity
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          protein dimerization activity
          peroxiredoxin activity
          peroxynitrite reductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P137618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905416 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P137618 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P137618 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P137618 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P137618 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700523457 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56649 - -
          Agilent Rat cDNA Microarray (G4105A) 700523457 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728246 - -
          Affymetrix Rat Gene 1.0 ST Array 10728244 - -
          Affymetrix Rat Gene 1.0 ST Array 10728241 - -
          Affymetrix Rat Gene 1.0 ST Array 10728242 - -
          Affymetrix Rat Gene 1.0 ST Array 10728243 - -
          Affymetrix Rat Gene 1.0 ST Array 10728245 - -
          Affymetrix Rat Gene 1.1 ST Array 10728240 - -
          Rat Genome 230 2.0 Array 1367677_at - -
          Rat Genome 230 2.0 Array 1367677_PM_at - -
          Affymetrix HT Rat Focus Array 1367677_PM_at - -
          Affymetrix HT Rat Focus Array 1367677_at - -
          Affymetrix Rat Clariom S Assay PSR0100067239.rn.1 TC0100008151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100067245.rn.1 TC0100008151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100067238.rn.1 TC0100008151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100067244.rn.1 TC0100008151.rn.1 RTA TC
          Rat Expression Set 230 1367677_PM_at - -
          Rat Expression Set 230 1367677_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728240 - -
          Rat Genome U34 Set rc_AI105266_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21530556 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363325 - -
          RatRef-12 Expression BeadChip ILMN_64677 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline