Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586086_m1
          See other NUP88 GE Assays ›
          Gene Symbol:
          Nup88
          Gene Name:
          nucleoporin 88
          Gene Aliases:
          Nup84, Prei2
          Chromosome Location:
          Chr.10: 57581828 - 57606171 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586086_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nup88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053616.1 NP_446068.1 9 - 10 1403 145
          GenBank mRNA BC072524.1 - 9 - 10 1397 7097375 145
          U93692.1 - 9 - 10 1403 145

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003146.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nup88

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s)

          protein transport
          mRNA transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P522336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183875 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99703 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141632 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P522336 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P522336 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P522336 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P522336 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700370786 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40290 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25874 - -
          Agilent Rat cDNA Microarray (G4105A) 700370786 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744560 - -
          Affymetrix Rat Gene 1.0 ST Array 10744551 - -
          Affymetrix Rat Gene 1.0 ST Array 10744557 - -
          Affymetrix Rat Gene 1.0 ST Array 10744556 - -
          Affymetrix Rat Gene 1.0 ST Array 10744566 - -
          Affymetrix Rat Gene 1.0 ST Array 10744554 - -
          Affymetrix Rat Gene 1.0 ST Array 10744555 - -
          Affymetrix Rat Gene 1.0 ST Array 10744563 - -
          Affymetrix Rat Gene 1.0 ST Array 10744561 - -
          Affymetrix Rat Gene 1.0 ST Array 10744562 - -
          Affymetrix Rat Gene 1.0 ST Array 10744559 - -
          Affymetrix Rat Gene 1.0 ST Array 10744567 - -
          Affymetrix Rat Gene 1.0 ST Array 10744553 - -
          Affymetrix Rat Gene 1.0 ST Array 10744552 - -
          Affymetrix Rat Gene 1.0 ST Array 10744565 - -
          Affymetrix Rat Gene 1.0 ST Array 10744564 - -
          Affymetrix Rat Gene 1.0 ST Array 10744558 - -
          Affymetrix Rat Gene 1.1 ST Array 10744550 - -
          Rat Genome 230 2.0 Array 1368199_at - -
          Rat Genome 230 2.0 Array 1368199_PM_at - -
          Affymetrix HT Rat Focus Array 1368199_PM_at - -
          Affymetrix HT Rat Focus Array 1368199_at - -
          Affymetrix Rat Clariom S Assay PSR1000028325.rn.1 TC1000003146.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000028323.rn.1 TC1000003146.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000028327.rn.1 TC1000003146.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000028324.rn.1 TC1000003146.rn.1 RTA TC
          Rat Expression Set 230 1368199_PM_at - -
          Rat Expression Set 230 1368199_at - -
          Affymetrix Rat Gene 1.0 ST Array 10744550 - -
          Rat Genome U34 Set U93692_at - -
          Rat Genome U34 Set AA799429_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20732216 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358895 - -
          RatRef-12 Expression BeadChip ILMN_51981 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline