Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586092_m1
          See other CPB2 GE Assays ›
          Gene Symbol:
          Cpb2
          Gene Name:
          carboxypeptidase B2 (plasma)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 57290849 - 57339762 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586092_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cpb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053617.2 NP_446069.1 9 - 10 1024 102
          GenBank mRNA BC091133.1 - 9 - 10 988 102
          BC107447.1 - 9 - 10 1017 7323668 102
          FQ210930.1 - 3 - 4 336 102
          FQ218610.1 - 9 - 10 1030 102
          FQ219505.1 - 7 - 8 810 102
          FQ219645.1 - 9 - 10 1096 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000843.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cpb2

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of extracellular matrix constituent secretion
          proteolysis
          blood coagulation
          response to heat
          negative regulation of plasminogen activation
          response to drug
          fibrinolysis
          negative regulation of fibrinolysis
          cellular response to glucose stimulus
          liver regeneration
          negative regulation of hepatocyte proliferation
          carboxypeptidase activity
          metallocarboxypeptidase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P557487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391378 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8087 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48640 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P557487 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P557487 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P557487 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P557487 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72120 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16382 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72923 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781522 - -
          Affymetrix Rat Gene 1.0 ST Array 10781517 - -
          Affymetrix Rat Gene 1.0 ST Array 10781524 - -
          Affymetrix Rat Gene 1.0 ST Array 10781518 - -
          Affymetrix Rat Gene 1.0 ST Array 10781520 - -
          Affymetrix Rat Gene 1.0 ST Array 10781514 - -
          Affymetrix Rat Gene 1.0 ST Array 10781521 - -
          Affymetrix Rat Gene 1.0 ST Array 10781515 - -
          Affymetrix Rat Gene 1.0 ST Array 10781519 - -
          Affymetrix Rat Gene 1.0 ST Array 10781516 - -
          Affymetrix Rat Gene 1.0 ST Array 10781523 - -
          Affymetrix Rat Gene 1.1 ST Array 10781513 - -
          Rat Genome 230 2.0 Array 1369206_PM_at - -
          Rat Genome 230 2.0 Array 1369206_at - -
          Affymetrix HT Rat Focus Array 1369206_at - -
          Affymetrix HT Rat Focus Array 1369206_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500005807.rn.1 TC1500000843.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005805.rn.1 TC1500000843.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005806.rn.1 TC1500000843.rn.1 RTA TC
          Rat Expression Set 230 1369206_at - -
          Rat Expression Set 230 1369206_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781513 - -
          Rat Genome U34 Set rc_AA998857_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22136357 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357330 - -
          RatRef-12 Expression BeadChip ILMN_68325 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0