Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586119_m1
          See other CDC42BPB GE Assays ›
          Gene Symbol:
          Cdc42bpb
          Gene Name:
          CDC42 binding protein kinase beta (DMPK-like)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 135746743 - 135830143 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586119_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdc42bpb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053620.3 NP_446072.2 1 - 2 265 76
          XM_006240562.2 XP_006240624.1 1 - 2 414 76
          GenBank mRNA AF021936.1 - 1 - 2 225 76
          AY277590.1 - 1 - 2 265 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003208.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdc42bpb

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          cell migration
          actin cytoskeleton organization
          actomyosin structure organization
          actin cytoskeleton reorganization
          intracellular signal transduction
          regulation of catalytic activity
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          Rho GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P081597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83075 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37366 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7891 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167593 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138516 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72040 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081597 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081597 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P081597 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12876 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P104590 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11098 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46521 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95019 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62492 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46358 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78512 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3819 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10892248 - -
          Affymetrix Rat Gene 1.0 ST Array 10892250 - -
          Affymetrix Rat Gene 1.0 ST Array 10892257 - -
          Affymetrix Rat Gene 1.0 ST Array 10892242 - -
          Affymetrix Rat Gene 1.0 ST Array 10892231 - -
          Affymetrix Rat Gene 1.0 ST Array 10892237 - -
          Affymetrix Rat Gene 1.0 ST Array 10892246 - -
          Affymetrix Rat Gene 1.0 ST Array 10892249 - -
          Affymetrix Rat Gene 1.0 ST Array 10892244 - -
          Affymetrix Rat Gene 1.0 ST Array 10892225 - -
          Affymetrix Rat Gene 1.0 ST Array 10892255 - -
          Affymetrix Rat Gene 1.0 ST Array 10892245 - -
          Affymetrix Rat Gene 1.0 ST Array 10892228 - -
          Affymetrix Rat Gene 1.0 ST Array 10892230 - -
          Affymetrix Rat Gene 1.0 ST Array 10892253 - -
          Affymetrix Rat Gene 1.0 ST Array 10892251 - -
          Affymetrix Rat Gene 1.0 ST Array 10892234 - -
          Affymetrix Rat Gene 1.0 ST Array 10892232 - -
          Affymetrix Rat Gene 1.0 ST Array 10892254 - -
          Affymetrix Rat Gene 1.0 ST Array 10892258 - -
          Affymetrix Rat Gene 1.0 ST Array 10892229 - -
          Affymetrix Rat Gene 1.0 ST Array 10892240 - -
          Affymetrix Rat Gene 1.0 ST Array 10892238 - -
          Affymetrix Rat Gene 1.0 ST Array 10892239 - -
          Affymetrix Rat Gene 1.0 ST Array 10892223 - -
          Affymetrix Rat Gene 1.0 ST Array 10892233 - -
          Affymetrix Rat Gene 1.0 ST Array 10892256 - -
          Affymetrix Rat Gene 1.0 ST Array 10892241 - -
          Affymetrix Rat Gene 1.0 ST Array 10892226 - -
          Affymetrix Rat Gene 1.0 ST Array 10892236 - -
          Affymetrix Rat Gene 1.0 ST Array 10892260 - -
          Affymetrix Rat Gene 1.0 ST Array 10892247 - -
          Affymetrix Rat Gene 1.0 ST Array 10892259 - -
          Affymetrix Rat Gene 1.0 ST Array 10892252 - -
          Affymetrix Rat Gene 1.0 ST Array 10892224 - -
          Affymetrix Rat Gene 1.0 ST Array 10892227 - -
          Affymetrix Rat Gene 1.0 ST Array 10892235 - -
          Affymetrix Rat Gene 1.0 ST Array 10892243 - -
          Affymetrix Rat Gene 1.1 ST Array 10892222 - -
          Rat Genome 230 2.0 Array 1387695_PM_at - -
          Rat Genome 230 2.0 Array 1372018_PM_at - -
          Rat Genome 230 2.0 Array 1387695_at - -
          Rat Genome 230 2.0 Array 1372018_at - -
          Affymetrix HT Rat Focus Array 1372018_at - -
          Affymetrix HT Rat Focus Array 1387695_at - -
          Affymetrix HT Rat Focus Array 1387695_PM_at - -
          Affymetrix HT Rat Focus Array 1372018_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600022733.rn.1 TC0600003208.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022731.rn.1 TC0600003208.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022732.rn.1 TC0600003208.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600022734.rn.1 TC0600003208.rn.1 RTA TC
          Rat Expression Set 230 1372018_at - -
          Rat Expression Set 230 1387695_at - -
          Rat Expression Set 230 1387695_PM_at - -
          Rat Expression Set 230 1372018_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10892222 - -
          Rat Genome U34 Set AF021936_at - -
          Rat Genome U34 Set rc_AA818720_at - -
          Rat Genome U34 Set AF021936_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22273554 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370386 - -
          RatRef-12 Expression BeadChip ILMN_53263 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline