Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586167_m1
          See other POM121 GE Assays ›
          Gene Symbol:
          Pom121
          Gene Name:
          POM121 transmembrane nucleoporin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 24324590 - 24341943 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586167_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pom121
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053622.1 NP_446074.1 3 - 4 953 59
          XM_008769111.1 XP_008767333.1 3 - 4 1442 59
          GenBank mRNA Z21513.1 - 3 - 4 953 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000424.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pom121

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear pore organization
          protein transport
          mRNA transport
          structural constituent of nuclear pore

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P427560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84443 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P427560 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P427560 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P427560 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P427560 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50759 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62321 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757681 - -
          Affymetrix Rat Gene 1.0 ST Array 10757686 - -
          Affymetrix Rat Gene 1.0 ST Array 10757682 - -
          Affymetrix Rat Gene 1.0 ST Array 10757687 - -
          Affymetrix Rat Gene 1.0 ST Array 10757680 - -
          Affymetrix Rat Gene 1.0 ST Array 10757685 - -
          Affymetrix Rat Gene 1.0 ST Array 10757676 - -
          Affymetrix Rat Gene 1.0 ST Array 10757679 - -
          Affymetrix Rat Gene 1.0 ST Array 10757678 - -
          Affymetrix Rat Gene 1.0 ST Array 10757675 - -
          Affymetrix Rat Gene 1.0 ST Array 10757684 - -
          Affymetrix Rat Gene 1.0 ST Array 10757677 - -
          Affymetrix Rat Gene 1.0 ST Array 10757683 - -
          Affymetrix Rat Gene 1.1 ST Array 10757674 - -
          Rat Genome 230 2.0 Array 1386957_at - -
          Rat Genome 230 2.0 Array 1386957_PM_at - -
          Affymetrix HT Rat Focus Array 1386957_at - -
          Affymetrix HT Rat Focus Array 1386957_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200003773.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003781.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003783.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003785.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003789.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003776.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003784.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003788.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003790.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003774.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003777.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003782.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003787.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003778.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003779.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003780.rn.1 TC1200000424.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003786.rn.1 TC1200000424.rn.1 RTA TC
          Rat Expression Set 230 1386957_at - -
          Rat Expression Set 230 1386957_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757674 - -
          Rat Genome U34 Set rc_AI178208_at - -
          Rat Genome U34 Set rc_AA925591_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20896792 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365360 - -
          RatRef-12 Expression BeadChip ILMN_48593 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline