Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586179_m1
          See other ACSL4 GE Assays ›
          Gene Symbol:
          Acsl4
          Gene Name:
          acyl-CoA synthetase long-chain family member 4
          Gene Aliases:
          Acs4, Facl4
          Chromosome Location:
          Chr.X: 113596247 - 113660024 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586179_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acsl4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053623.1 NP_446075.1 15 - 16 2042 99
          XM_006257314.2 XP_006257376.1 15 - 16 2380 99
          XM_006257315.2 XP_006257377.1 16 - 17 2161 99
          XM_006257316.2 XP_006257378.1 15 - 16 2185 99
          XM_008773385.1 XP_008771607.1 16 - 17 2454 99
          GenBank mRNA D85189.1 - 15 - 16 2042 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002274.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acsl4

          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          long-chain fatty acid metabolic process
          lipid metabolic process
          fatty acid metabolic process
          triglyceride metabolic process
          response to nutrient
          lipid biosynthetic process
          fatty acid transport
          neuron differentiation
          positive regulation of cell growth
          negative regulation of prostaglandin secretion
          embryonic process involved in female pregnancy
          dendritic spine development
          response to interleukin-15
          long-chain fatty acid-CoA ligase activity
          ATP binding
          very long-chain fatty acid-CoA ligase activity
          arachidonate-CoA ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P654061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863131 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155996 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5818 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8516 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46467 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P654061 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P654061 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P654061 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P654061 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P375004 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700373919 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15832 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51885 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81584 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63600 - -
          Agilent Rat cDNA Microarray (G4105A) 702211677 - -
          Agilent Rat cDNA Microarray (G4105A) 700373919 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10932801 - -
          Affymetrix Rat Gene 1.0 ST Array 10932809 - -
          Affymetrix Rat Gene 1.0 ST Array 10932797 - -
          Affymetrix Rat Gene 1.0 ST Array 10932808 - -
          Affymetrix Rat Gene 1.0 ST Array 10932796 - -
          Affymetrix Rat Gene 1.0 ST Array 10932811 - -
          Affymetrix Rat Gene 1.0 ST Array 10932802 - -
          Affymetrix Rat Gene 1.0 ST Array 10932799 - -
          Affymetrix Rat Gene 1.0 ST Array 10932806 - -
          Affymetrix Rat Gene 1.0 ST Array 10932803 - -
          Affymetrix Rat Gene 1.0 ST Array 10932807 - -
          Affymetrix Rat Gene 1.0 ST Array 10932805 - -
          Affymetrix Rat Gene 1.0 ST Array 10932812 - -
          Affymetrix Rat Gene 1.0 ST Array 10932800 - -
          Affymetrix Rat Gene 1.0 ST Array 10932810 - -
          Affymetrix Rat Gene 1.0 ST Array 10932804 - -
          Affymetrix Rat Gene 1.1 ST Array 10932795 - -
          Rat Genome 230 2.0 Array 1387101_PM_at - -
          Rat Genome 230 2.0 Array 1387101_at - -
          Affymetrix HT Rat Focus Array 1387101_at - -
          Affymetrix HT Rat Focus Array 1387101_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00014251.rn.1 TC0X00002274.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00014249.rn.1 TC0X00002274.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00014250.rn.1 TC0X00002274.rn.1 RTA TC
          Rat Expression Set 230 1387101_PM_at - -
          Rat Expression Set 230 1387101_at - -
          Affymetrix Rat Gene 1.0 ST Array 10932795 - -
          Rat Genome U34 Set D85189_at - -
          Rat Genome U34 Set rc_AI236284_s_at - -
          Rat Genome U34 Set rc_AA925424_at - -
          Rat Genome U34 Set rc_AI045002_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22340085 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371034 - -
          RatRef-12 Expression BeadChip ILMN_68834 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline